Skip to Content
Merck
All Photos(1)

Key Documents

1249406

USP

Escitalopram oxalate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

S-(+)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate, S-(+)-Citalopram oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21FN2O · C2H2O4
CAS Number:
Molecular Weight:
414.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

escitalopram, escitalopram

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.CN(C)CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

InChI

1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m0./s1

InChI key

KTGRHKOEFSJQNS-BDQAORGHSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the authority of the issuing pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Escitalopram oxalate, the S-enantiomer of racemic citalopram, belongs to the class of compounds known as antidepressants, that works as a selective serotonin-reuptake inhibitor (SSRI).

Application

Escitalopram oxalate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Escitalopram Oral Solution
  • Escitalopram Oxalate
  • Escitalopram Tablets

Biochem/physiol Actions

Escitalopram is a selective serotonin reuptake inhibitor (SSRI), the S-enantiomer and eutomer of citalopram.
Escitalopram oxalate is a selective serotonin reuptake inhibitor (SSRI); the S-enantiomer and eutomer of citalopram.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric and reversed-phase high-performance liquid chromatographic methods for simultaneous determination of escitalopram oxalate and clonazepam in combined tablet dosage form
Gandhi VS, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 91(1), 33-38 (2008)
Application Of Stability Indicating Hptlc Method For Quantitative Determination Of Escitalopram Oxalate In Pharmaceutical Dosage Form.
Mahadik VM, et al.
Eurasian Journal of Analytical Chemistry, 2(2) (2007)
Simultaneous HPTLC determination of escitalopram oxalate and clonazepam in combined tablets.
Dhavale N, et al.
Chromatographia, 67(5-6), 487-490 (2008)
Kristan A Leech et al.
Journal of neurotrauma, 31(15), 1334-1342 (2014-04-20)
Incomplete spinal cord injury (iSCI) often results in significant motor impairments that lead to decreased functional mobility. Loss of descending serotonergic (5HT) input to spinal circuits is thought to contribute to motor impairments, with enhanced motor function demonstrated through augmentation
Dietmar Hestermann et al.
Behavioural brain research, 273, 155-165 (2014-07-22)
Serotonergic (5-HT) drugs are widely used in the clinical management of mood and anxiety disorders. However, it is reported that acute 5-HT treatment elicits anxiogenic-like behavior. Interestingly, the periaqueductal gray (PAG), a midbrain structure which regulates anxiety behavior - has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service