Skip to Content
Merck
All Photos(1)

Key Documents

E4786

Sigma-Aldrich

Escitalopram oxalate

≥98% (HPLC), powder, serotonin reuptake inhibitor

Synonym(s):

S-(+)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile oxalate, S-(+)-Citalopram oxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21FN2O · C2H2O4
CAS Number:
Molecular Weight:
414.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Escitalopram oxalate, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥15 mg/mL

originator

Forest Labs

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.CN(C)CCC[C@]1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

InChI

1S/C20H21FN2O.C2H2O4/c1-23(2)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24-20;3-1(4)2(5)6/h4-9,12H,3,10-11,14H2,1-2H3;(H,3,4)(H,5,6)/t20-;/m0./s1

InChI key

KTGRHKOEFSJQNS-BDQAORGHSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

Escitalopram oxalate is an anti-depressant.[1] It is used to treat panic disorder[1] and major depressive disorder(MDD).[2]

Application

Escitalopram oxalate has been used as an antidepressant in rats exposed to chronic mild stress (CMS).[3] It has also been used for scintillation proximity-based thermostability assay.[4]

Biochem/physiol Actions

Escitalopram is a selective serotonin reuptake inhibitor (SSRI), the S-enantiomer and eutomer of citalopram.
Escitalopram oxalate is a selective serotonin reuptake inhibitor (SSRI); the S-enantiomer and eutomer of citalopram.

Features and Benefits

This compound was developed by Forest Labs. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermostabilization, expression, purification, and crystallization of the human serotonin transporter bound to S-citalopram
Coleman JA, et al.
Journal of Visualized Experiments, e54792-e54792 (2016)
Colorimetric method for the estimation of escitalopram oxalate in tablet dosage form
Vetrichelvan T, et al.
Indian Journal of Pharmaceutical Sciences, 72(2), 269-269 (2010)
André Russowsky Brunoni et al.
Sao Paulo medical journal = Revista paulista de medicina, 133(3), 252-263 (2015-07-16)
Major depressive disorder (MDD) is a common psychiatric condition, mostly treated with antidepressant drugs, which are limited due to refractoriness and adverse effects. We describe the study rationale and design of ELECT-TDCS (Escitalopram versus Electric Current Therapy for Treating Depression
Escitalopram
Waugh J and Goa KL
CNS Drugs, 17(5), 343-362 (2003)
Modulation of the antioxidant nuclear factor (erythroid 2-derived)-like 2 pathway by antidepressants in rats
Martin-Hernandez D, et al.
Neuropharmacology, 103, 79-91 (2016)

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service