Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
aminosalicylic acid
manufacturer/tradename
USP
mp
135-145 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
Nc1ccc(C(O)=O)c(O)c1
InChI
1S/C7H7NO3/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,9H,8H2,(H,10,11)
InChI key
WUBBRNOQWQTFEX-UHFFFAOYSA-N
General description
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| mp 135-145 °C (lit.) | mp 158-161 °C (lit.) | mp 213-217 °C (lit.) | mp >300 °C (lit.) |
| API family aminosalicylic acid | API family salicylic acid | API family salicylic acid | API family mesalazine, sulfadimethoxine, sulfadiazine |
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



