Skip to Content
Merck
All Photos(1)

Key Documents

1005004

USP

Acetazolamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3S2
CAS Number:
Molecular Weight:
222.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

acetazolamide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)Nc1nnc(s1)S(N)(=O)=O

InChI

1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)

InChI key

BZKPWHYZMXOIDC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetazolamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Acetazolamide Compounded Oral Suspension
  • Acetazolamide Extended-Release Capsules
  • Acetazolamide for Injection
  • Acetazolamide Tablets
Carbonic anhydrase inhibitor; increases cerebral blood flow.

Biochem/physiol Actions

Inhibits water permeability of membranes by interacting with aquaporins

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil D Ritchie et al.
Journal of travel medicine, 19(5), 298-307 (2012-09-05)
Acetazolamide has been reported to be effective in the prevention of acute mountain sickness (AMS). Our aim was to conduct a systematic review of randomized, placebo-controlled trials of acetazolamide in the prevention of AMS. Studies were identified by searching the
Bengt Kayser et al.
High altitude medicine & biology, 13(2), 82-92 (2012-06-26)
Acetazolamide is used to prevent acute mountain sickness (AMS). We assessed efficacy and harm of acetazolamide for the prevention of AMS, and tested for dose-responsiveness. We systematically searched electronic databases (until April 2011) for randomized trials comparing acetazolamide with placebo
Emma V Low et al.
BMJ (Clinical research ed.), 345, e6779-e6779 (2012-10-20)
To assess the efficacy of three different daily doses of acetazolamide in the prevention of acute mountain sickness and to determine the lowest effective dose. Systematic review and meta-analysis. Medline and Embase along with a hand search of selected bibliographies.
A S Vagal et al.
AJNR. American journal of neuroradiology, 30(5), 876-884 (2009-02-28)
The acetazolamide (ACZ) challenge test is a useful clinical tool and a reliable predictor of critically reduced perfusion. In patients with chronic steno-occlusive disease, the ability to maintain normal cerebral blood flow by reducing vascular resistance secondary to autoregulatory vasodilation
David E Leaf et al.
Journal of applied physiology (Bethesda, Md. : 1985), 102(4), 1313-1322 (2006-10-07)
Acetazolamide, a potent carbonic anhydrase (CA) inhibitor, is the most commonly used and best-studied agent for the amelioration of acute mountain sickness (AMS). The actual mechanisms by which acetazolamide reduces symptoms of AMS, however, remain unclear. Traditionally, acetazolamide's efficacy has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service