Skip to Content
Merck
All Photos(1)

Key Documents

Y0001084

Acetazolamide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acetazolamide, 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3S2
CAS Number:
Molecular Weight:
222.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

acetazolamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)Nc1nnc(s1)S(N)(=O)=O

InChI

1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)

InChI key

BZKPWHYZMXOIDC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GBL305118GBL305108GBL305384
manufacturer/tradename

Grace Bio-Labs 305177

manufacturer/tradename

Grace Bio-Labs 305118

manufacturer/tradename

Grace Bio-Labs 305108

manufacturer/tradename

Grace Bio-Labs 305384

packaging

pack of 20 ea

packaging

pack of 20 ea

packaging

pack of 20 ea

packaging

pack of 1 ea

feature

1 (Pads)

feature

8 (Pads)

feature

8 (Pads)

feature

384 (Pads)

L × W

60 mm × 20 mm , Pads

L × W

20 mm × 6 mm , Pads

L × W

15 mm × 6 mm , Pads

L × W

2.5 mm × 2.5 mm , Pads

binding type

high binding surface

binding type

high binding surface

binding type

high binding surface

binding type

high binding surface

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbonic anhydrase inhibitor; increases cerebral blood flow.

Biochem/physiol Actions

Inhibits water permeability of membranes by interacting with aquaporins

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bengt Kayser et al.
High altitude medicine & biology, 13(2), 82-92 (2012-06-26)
Acetazolamide is used to prevent acute mountain sickness (AMS). We assessed efficacy and harm of acetazolamide for the prevention of AMS, and tested for dose-responsiveness. We systematically searched electronic databases (until April 2011) for randomized trials comparing acetazolamide with placebo
Emma V Low et al.
BMJ (Clinical research ed.), 345, e6779-e6779 (2012-10-20)
To assess the efficacy of three different daily doses of acetazolamide in the prevention of acute mountain sickness and to determine the lowest effective dose. Systematic review and meta-analysis. Medline and Embase along with a hand search of selected bibliographies.
A S Vagal et al.
AJNR. American journal of neuroradiology, 30(5), 876-884 (2009-02-28)
The acetazolamide (ACZ) challenge test is a useful clinical tool and a reliable predictor of critically reduced perfusion. In patients with chronic steno-occlusive disease, the ability to maintain normal cerebral blood flow by reducing vascular resistance secondary to autoregulatory vasodilation
Neil D Ritchie et al.
Journal of travel medicine, 19(5), 298-307 (2012-09-05)
Acetazolamide has been reported to be effective in the prevention of acute mountain sickness (AMS). Our aim was to conduct a systematic review of randomized, placebo-controlled trials of acetazolamide in the prevention of AMS. Studies were identified by searching the
Indu Pal Kaur et al.
International journal of pharmaceutics, 248(1-2), 1-14 (2002-11-14)
Through this review it is contemplated that acetazolamide (ACZ), an age-old treatment for glaucoma with a myriad of side effects and inadequate topical effectiveness, may be formulated into a topically effective agent by utilizing various newer formulation approaches of ocular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service