PHL86134
Quercetin 3-robinoside 7-glucoside
phyproof® Reference Substance
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-robinoside 7-glucoside, Quercetin 3-rhamnogalactoside 7-glucoside
About This Item
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
manufacturer/tradename
PhytoLab
format
neat
SMILES string
O=C1C2=C(O)C=C(O[C@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O3)C=C2OC(C4=CC=C(O)C(O)=C4)=C1O[C@H]5[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO[C@H]6[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O6)O5
InChI
1S/C33H40O21/c1-9-19(38)23(42)26(45)31(49-9)48-8-17-21(40)25(44)28(47)33(53-17)54-30-22(41)18-14(37)5-11(50-32-27(46)24(43)20(39)16(7-34)52-32)6-15(18)51-29(30)10-2-3-12(35)13(36)4-10/h2-6,9,16-17,19-21,23-28,31-40,42-47H,7-8H2,1H3/t9-,16+,17+,19-,20+,21-
InChI key
SPUFXPFDJYNCFD-MSLGMOTCSA-N
Related Categories
General description
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service