Skip to Content
Merck
All Photos(1)

Key Documents

MSCAL5

Supelco

ProteoMass LTQ/FT-Hybrid ESI Pos. Mode Cal Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

product line

ProteoMass

CofA

current certificate can be downloaded

packaging

pkg of 1 × 10 mL

storage condition

protect from light

concentration

(varied conc.)

technique(s)

mass spectrometry (MS): suitable

format

multi-component solution

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The ProteoMass ESI Calibration Kit for LTQ based Hybrids, positive ion mode calibration solution, enables calibration of Thermo Scientific hybrid instruments equipped with ESI source in positive ion mode. It covers a mass range from m/z 138 to m/z 1822, and is therefore usable for calibrations between m/z 50 and 2000.

Application

ProteoMass LTQ/FT-hybrid ESI Pos. mode cal mix has been used as a calibration standard for the mass spectrometric analysis of serum samples of patients affected by mugwort pollen allergy induced by pollen extract of Artemisia vulgaris.

Features and Benefits

Discover the Advantages for Yourself!
  • Ready-to-use
  • Developed and qualified specifically for use with the LTQ based Thermo Scientific hybrid instruments (LTQ FT, LTQ FT Ultra, and LTQ Orbitrap series) with ESI source and Thermo Scientific Exactive
  • Conveniently packaged, freeing you from time consuming mixing and dilution steps, allowing you to focus on the acquistion of quality data

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.1 °F - closed cup

Flash Point(C)

5.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Apostolovic et al.
Scientific reports, 7(1), 5201-5201 (2017-07-14)
The mammalian carbohydrate galactose-α1,3-galactose (α-Gal) causes a novel form of food allergy, red meat allergy, where patients experience severe allergic reactions several hours after red meat consumption. Here we explored gastric digestion of α-Gal glycoproteins using an in vitro model.
Hypoallergenic acid-sensitive modification preserves major mugwort allergen fold and delivers full repertoire of MHC class II-binding peptides during endolysosomal degradation
Stanic-Vucinic D, et al.
Royal Society of Chemistry Advances, 91(6), 88216-88228 (2016)
Adam P Bateman et al.
Analytical chemistry, 82(19), 8010-8016 (2010-09-03)
This work demonstrates the utility of a particle-into-liquid sampler (PILS), a technique traditionally used for identification of inorganic ions present in ambient or laboratory aerosols, for the analysis of water-soluble organic aerosol (OA) using high-resolution electrospray ionization mass spectrometry (HR-ESI-MS).
Robert J Strife et al.
Rapid communications in mass spectrometry : RCM, 24(11), 1497-1501 (2010-05-21)
Kendrick Mass Mapping of complex sample compositions, via data derived by Fourier transform ion cyclotron resonance mass spectrometry (FTICRMS) at 1,000,000 resolution, is becoming more common. The Orbitrap mass spectrometer (Fourier Transform Axially Harmonic Orbital Trapping) also uses FT methods
Toine F H Bovee et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 33(5), 749-759 (2016-04-20)
In 2013 the Dutch authorities issued a warning against a dietary supplement that was linked to 11 reported adverse reactions, including heart problems and in one case even a cardiac arrest. In the UK a 20-year-old woman, said to have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service