Skip to Content
Merck
All Photos(1)

Documents

EC10A-1KT

Supelco

Fatty Acid Kit

(individually packaged, quantities indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Assay

99-100% (GLC, TLC)

CofA

current certificate can be downloaded

packaging

pkg of 1 kit

concentration

(individually packaged, quantities indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

carboxylic acid
ester

storage temp.

2-8°C

General description

A kit of even-carbon fatty acids. Each component individually packaged at quantities indicated.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Arachidic acid 5 g
    Behenic acid 5 g
    Decanoic acid 10 g
    Dodecanoic acid 10 g
    Hexanoic acid 10 mL
    Lignoceric acid 1 g
    Myristic acid 10 g
    Octanoic acid 10 mL
    Palmitic acid 10 g
    Stearic acid 5 g
    See All (10)

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianwen Wei et al.
The Journal of clinical investigation, 124(4), 1-13 (2014-03-20)
Insulin signaling in osteoblasts has been shown recently to contribute to whole-body glucose homeostasis in animals fed a normal diet; however, it is unknown whether bone contributes to the insulin resistance that develops in animals challenged by a high-fat diet
Feng Wang et al.
Reproduction (Cambridge, England), 153(2), 175-185 (2016-12-07)
Hormone-sensitive lipase-knockout (HSL-/-) mice exhibit azoospermia for unclear reasons. To explore the basis of sterility, we performed the following three experiments. First, HSL protein distribution in the testis was determined. Next, transcriptome analyses were performed on the testes of three
Martha María Juárez-Rodríguez et al.
Frontiers in cellular and infection microbiology, 10, 597517-597517 (2021-02-16)
Blocking virulence is a promising alternative to counteract Pseudomonas aeruginosa infections. In this regard, the phenomenon of cell-cell communication by quorum sensing (QS) is an important anti-virulence target. In this field, fatty acids (FA) have gained notoriety for their role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service