Skip to Content
Merck
All Photos(1)

Key Documents

CRM05139

Supelco

Ethiofencarb-sulfone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO4S
CAS Number:
Molecular Weight:
257.31
Beilstein:
1988595
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CCS(=O)(=O)Cc1ccccc1OC(=O)NC

InChI

1S/C11H15NO4S/c1-3-17(14,15)8-9-6-4-5-7-10(9)16-11(13)12-2/h4-7H,3,8H2,1-2H3,(H,12,13)

InChI key

IOPTXXRNXCPJGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Ethiofencarb sulfone is a metabolite of the carbamate insecticide, ethiofencarb, and an acetylcholinesterase inhibitor.
Its parent, ethiofencarb is not allowed for use in the European Union as per the commission regulation no 1107/2009, regarding the use of plant protection products and repealing the council directive 91/414/EEC.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
It can also be used as follows:

  • Monitoring of carbamate insecticides and their metabolites in green sea turtles on the Great Barrier Reef by Ultra high-performance liquid chromatography coupled to mass spectrometry
  • Multi-residue analysis of 250 pesticides by ultra-performance liquid chromatography (UHPLC) coupled to MS/MS in processed fruits
  • Simultaneous determination of 74 pesticide residues in orange juice samples using an optimized QuEChERS extraction and UHPLC-MS/MS
  • Development and validation of a UHPLC-MS/MS based method for the quantification of 31 carbamate pesticide residues in five different edible oil samples
  • Quantification of 166 pesticides at low ppb concentrations levels using UHPLC and electrospray ionization quadrupole Orbitrap high-resolution mass spectrometry (ESI Q-Orbitrap) along with QuECHERs in fruits and vegetables
  • Determination of 25 carbamate pesticide residues in different wine varieties using UHPLC-MS/MS for separation and detection and ultrasound-assisted surfactant-enhanced emulsification microextraction (UASEME) for sample treatment

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Moreno-González et al.
Journal of chromatography. A, 1315, 1-7 (2013-10-08)
A new sensitive multiresidue method based on ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) has been developed for the detection, confirmation and quantification of twenty five carbamates in wine samples. The separation was achieved in 5.5 min, using a Zorbax
Amy L Heffernan et al.
The Science of the total environment, 599-600, 1251-1262 (2017-05-20)
Chemical contamination poses a threat to ecosystem, biota and human health, and identifying these hazards is a complex challenge. Traditional hazard identification relies on a priori-defined targets of limited chemical scope, and is generally inappropriate for exploratory studies such as
Jian Wang et al.
Journal of agricultural and food chemistry, 60(49), 12088-12104 (2012-11-21)
This paper presents an application of ultrahigh-performance liquid chromatography and electrospray ionization quadrupole Orbitrap high-resolution mass spectrometry (UHPLC/ESI Q-Orbitrap) for determination of 166 pesticide residues in fruits and vegetables. Pesticides were extracted from the samples using the QuEChERS (quick, easy
Tiele M Rizzetti et al.
Food chemistry, 196, 25-33 (2015-11-26)
In this study, different extraction procedures based on the QuEChERS method were compared for the multiresidue determination of pesticides in orange juice by ultra high performance liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS). After choosing preliminary conditions, an experimental
David Moreno-González et al.
Talanta, 128, 299-304 (2014-07-26)
In this study a fast, selective and sensitive multiresidue method based on QuEChERS methodology has been evaluated and validated for the determination of carbamate pesticides, in edible vegetable oils by UHPLC-MS/MS. A new clean-up sorbent, Supel(TM) QuE Z-Sep(+), has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service