Skip to Content
Merck
All Photos(3)

Key Documents

289426

Sigma-Aldrich

4,4′−Dipyridyl

98%

Synonym(s):

4,4′-Bipyridyl, 4,4′-Bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2
CAS Number:
Molecular Weight:
156.18
Beilstein:
113176
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

bp

305 °C (lit.)

mp

109-112 °C (lit.)

SMILES string

c1cc(ccn1)-c2ccncc2

InChI

1S/C10H8N2/c1-5-11-6-2-9(1)10-3-7-12-8-4-10/h1-8H

InChI key

MWVTWFVJZLCBMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Dipyridyl (BPY) is an organic linker mainly used in the preparation of coordination polymers. It is a pyridine derivative in which pyridyl groups can rotate along the carbon-carbon structure. It can be used as a catalyst for photoelectrochemical reduction.
4,4′-bpy is popularly used to bridge metal centers to form coordination polymers with novel properties. 4,4′-bpy ligands can mediate electronic effects between paramagnetic metals ions by stabilizing the magnetic ordering.

Application

4,4′-Dipyridyl (4,4′-bipyridine, 4,4′-bpy) and Co (NCS)2 solutions were interdifussed to form single crystals Co(NCS)2(4,4′-bipy)2·2(CH3CH2)2O and Co(NCS)2(4,4′-bipy)2·2(CH3CH2)2O.
BPY is a ligand that forms a network between the transition metals which results in the formation of coordination bond. Hybrid nanosheets can be prepared by using a coordination polymer and BPY as a complex linker. It consists of two active sites which can be used for the development of drugs.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spin canting and metamagnetism in a 3D homometallic molecular material constructed by interpenetration of two kinds oc cobalt(II)-coordination polymer sheets
Zheng HM, et al.
Angewandte Chemie (International Edition in English), 44(20) (2005)
Coordination Polymers of Co(NCS)2 with Pyrazine and 4,4`-Bipyridine:? Syntheses and Structures
Lu J, et al.
Inorganic Chemistry, 36 (5), 923- 929 null
Bottom-up approach for the preparation of hybrid nanosheets based on coordination polymers made of metal-diethyloxaloacetate complexes linked by 4, 4′-bipyridine.
Portoles-Gil N, et al.
CrystEngComm, 19(34), 4972-4982 (2017)
Ultrafast photoinduced interfacial proton coupled electron transfer from cdse quantum dots to 4, 4′-bipyridine.
Chen J, et al.
Journal of the American Chemical Society, 138(3), 884-892 (2016)
Crystal engineering of coordination polymers using 4, 4′-bipyridine as a bond between transition metal atoms.
Biradha K, et al.
Chemical Communications (Cambridge, England), 138(40), 4169-4179 (2006)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service