Skip to Content
Merck
All Photos(1)

Key Documents

48994-U

Supelco

Residual Solvents in Packaging Material Mixture 1

certified reference material, 7.14 % (v/v), pkg of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

concentration

7.14 % (v/v)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-8°C

General description

Residual solvents from inks and dyes used in packaging graphics can impart off-flavors to food. In some cases, the residual solvents will penetrate the packaging material and be absorbed by the food product, compromising its quality. To ensure consumer safety and customer satisfaction, residual solvent standard mixtures aid food chemists in evaluating food packaging. All solvents are screened for identity and purity. A certificate of composition accompanies each standard.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    1-Butanol
    2-Butanol
    2-Butanone
    Butyl acetate
    Cyclohexane
    Cyclohexanone
    Ethanol
    2-Ethoxyethanol
    Ethyl acetate
    Isobutyl acetate
    Methanol
    Methyl acetate
    2-Methoxyethyl acetate
    Toluene
    See All (14)

Signal Word

Danger

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service