Skip to Content
Merck

1601281

USP

Residual Solvents Class 2 - Mixture A

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvents Class 2 - Mixture A USP reference standard for specified quality tests and assays.

Also used to prepare the standard stock solution for the screening of water-soluble and insoluble articles according to the United States Pharmacopeia (USP) chapters.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


wgk

WGK 3

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



{?467? RESIDUAL SOLVENTS
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 49(1), 6712-6712 (2022)



Global Trade Item Number

SKUGTIN
1601281-3X1.2ML04065271209237
1601281-3AMP04061838745064