Skip to Content
Merck
All Photos(1)

Key Documents

47848

Supelco

Potassium Sorbate

analytical standard

Synonym(s):

Sorbic acid potassium salt, 2,4-Hexadienoic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7KO2
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E202
Pricing and availability is not currently available.

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

fungi

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

[K+].C\C=C\C=C\C([O-])=O

InChI

1S/C6H8O2.K/c1-2-3-4-5-6(7)8;/h2-5H,1H3,(H,7,8);/q;+1/p-1/b3-2+,5-4+;

InChI key

CHHHXKFHOYLYRE-STWYSWDKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium sorbate is a food additive, used for the prevention of food spoilage originating from bacteria, molds or yeasts.[1][2] It exists in a variety of products such as pharmaceuticals, cosmetics and employed for preserving processed foods including fruit juices, soda, soy sauce, ketchup, etc.[1]
Potassium sorbate is a preservative[3], commonly used in a variety of food and beverages.[4] It is a food antimicrobial agent[5], which can act effectively along a wide pH range to inhibit the growth of yeast, molds and some bacteria.[4]

Application

Potassium sorbate may be used as an analytical reference standard for the quantification of the analyte in food, beverages[1] and soft drinks[2] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition effect of food preservatives on endoproteinases
Esimbekova.NE, et al.
Food Chemistry, 235, 294-297 (2017)
Development of reverse-phase high-performance liquid chromatography method for simultaneous determination of sodium benzoate and potassium sorbate in beverages
Pentanovska-Ilievska B, et al.
Acta Chromatographica , 29, 345-358 (2017)
A novel RP-HPLC method for simultaneous determination of potassium sorbate and sodium benzoate in soft drinks using C18-bonded monolithic silica column
Can ON, et al.
Journal of Separation Science, 34(16-17), 2214-2222 (2011)
Control of citrus postharvest green mold and sour rot by potassium sorbate combined with heat and fungicides
Smilanick.LJ, et al.
Postharvest biology and technology, 47, 226-238 (2008)
Ahmet C Gören et al.
Food chemistry, 175, 273-279 (2015-01-13)
High Performance Liquid Chromatography LC-UV and LC-MS/MS methods were developed and validated for quantitative analyses of sodium benzoate and potassium sorbate in foods and beverages. HPLC-UV and LC-MS/MS methods were compared for quantitative analyses of sodium benzoate and potassium sorbate

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service