Skip to Content
Merck
All Photos(1)

Documents

PHR1231

Supelco

Sodium benzoate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1055002
traceable to USP 1613564

API family

sodium benzoate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium benzoate is a sodium salt of benzoic acid, that is freely soluble in water compared to benzoic acid. It is generally used as an antimicrobial preservative in cosmetics, food, and pharmaceuticals.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Sodium benzoate may be used as a pharmaceutical reference standard for the determination of the analyte in raw material, food, cosmetics and pharmaceutical formulations by titrimetry and liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0283 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Benzoate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 883(1-2), 3566-3567 (2017)
HPLC and LC-MS/MS methods for determination of sodium benzoate and potassium sorbate in food and beverages: Performances of local accredited laboratories via proficiency tests in Turkey
Goren AC, et al
Food Chemistry, 175(3), 273-279 (2015)
High-pressure liquid chromatographic assay of dextromethorphan hydrobromide, guaifenesin, and sodium benzoate in an expectorant syrup
Chen TM, et al.
Journal of Chromatographic Science, 26(12), 636-639 (1988)
Simultaneous determination of aspartame, acesulfame-K, saccharin, citric acid and sodium benzoate in various food products using HPLC-CAD-UV/DAD
Grembecka M, et al
European Food Research and Technology, 238(3), 357-365 (2014)
Sodium Benzoate
Pharmacopeia, US
United States Pharmacopeia/National Formulary, 38(3)(1-2), 5951-5951 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service