Skip to Content
Merck
All Photos(1)

Key Documents

442498

Supelco

Bromochloromethane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2Cl
CAS Number:
Molecular Weight:
129.38
Beilstein:
1730801
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor density

4.5 (vs air)

vapor pressure

117 mmHg ( 20 °C)

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.482 (lit.)

bp

68 °C (lit.)

mp

−88 °C (lit.)

density

1.991 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

ClCBr

InChI

1S/CH2BrCl/c2-1-3/h1H2

InChI key

JPOXNPPZZKNXOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers bromochloromethane for a broad-range applications such as environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Application

Bromochloromethane has been used in form of an internal standard mixture, for the identification and separation of volatile organic compounds emitted from several sources, using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Ozone 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile Profiles of Emissions from Different Activities Analyzed Using Canister Samplers and Gas Chromatography-Mass Spectrometry (GC/MS) Analysis: A Case Study
Orecchio S, et al.
International Journal of Environmental Research and Public Health, 14, 195-195 (2017)
L Abecia et al.
Journal of dairy science, 95(4), 2027-2036 (2012-03-31)
Several technologies have been tested to reduce enteric methanogenesis, but very few have been successfully used in practical conditions for livestock. Furthermore, the consequences of reduced rumen methane production on animal performance and milk quality are poorly understood. The aim
D Pankow et al.
Polish journal of occupational medicine and environmental health, 5(3), 245-250 (1992-01-01)
An oral dose of 6.2 mmoles of diachloromethane (DCM), bromochloromethane (BCM) or dibromomethane (DBM) per kg body mass yielded a maximum carboxyhemoglobin (COHb) level of about 9% (at 6 hr), 11% (at 8 hr) and 22% (at 12 hr), respectively.
Stuart E Denman et al.
FEMS microbiology ecology, 62(3), 313-322 (2007-10-24)
Methyl coenzyme-M reductase A (mcrA) clone libraries were generated from microbial DNA extracted from the rumen of cattle fed a roughage diet with and without supplementation of the antimethanogenic compound bromochloromethane. Bromochloromethane reduced total methane emissions by c. 30%, with
M L Gargas et al.
Toxicology and applied pharmacology, 86(3), 341-352 (1986-12-01)
In vivo metabolic constants were determined in male Fischer rats for five chemicals: 1,1-dichloroethylene (1,1-DCE), diethyl ether (DE), bromochloromethane (BCM), methyl chloroform (MC), and carbon tetrachloride (CCl4). A closed recirculated exposure system was used to collect a series of uptake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service