Skip to Content
Merck
All Photos(2)

Key Documents

139181

Sigma-Aldrich

Bromodichloromethane

≥97%

Synonym(s):

Dichlorobromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCHCl2
CAS Number:
Molecular Weight:
163.83
Beilstein:
1697005
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

contains

potassium carbonate as stabilizer

refractive index

n20/D 1.497 (lit.)

bp

87 °C (lit.)

mp

−55 °C (lit.)

solubility

water: insoluble

density

1.98 g/mL at 25 °C (lit.)

functional group

bromo
chloro

SMILES string

ClC(Cl)Br

InChI

1S/CHBrCl2/c2-1(3)4/h1H

InChI key

FMWLUWPQPKEARP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bromodichloromethane was used to investigate the in vivo genotoxicity of chlorodibromomethane, bromodichloromethane and bromoform in mice.

Biochem/physiol Actions

Bromodichloromethane (BDCM) induces steatohepatitic injury in human liver. BDCM induces the cell proliferation in different tissues and kidney damage by renal tuble cell proliferation in male F344 rats.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Stocker et al.
Mutagenesis, 12(3), 169-173 (1997-05-01)
Chlorination of drinking water results in the formation of chlorodibromomethane, bromodichloromethane and bromoform. These trihalomethanes have all shown evidence of genotoxicity in bacterial and mammalian cell systems in vitro and some evidence of carcinogenicity in rodents. Chlorodibromomethane and bromodichloromethane have
Teresa L Leavens et al.
Toxicological sciences : an official journal of the Society of Toxicology, 99(2), 432-445 (2007-07-28)
Exposure to bromodichloromethane (BDCM), one of the most prevalent disinfection byproducts in drinking water, can occur via ingestion of water and by dermal absorption and inhalation during activities such as bathing and showering. The objectives of this research were to
Suvarthi Das et al.
Toxicology and applied pharmacology, 269(3), 297-306 (2013-02-27)
Today's developed world faces a major public health challenge in the rise in the obese population and the increased incidence in fatty liver disease. There is a strong association among diet induced obesity, fatty liver disease and development of nonalcoholic
Suvarthi Das et al.
American journal of physiology. Gastrointestinal and liver physiology, 305(12), G950-G963 (2013-10-26)
Recent studies indicate that metabolic oxidative stress, autophagy, and inflammation are hallmarks of nonalcoholic steatohepatitis (NASH) progression. However, the molecular mechanisms that link these important events in NASH remain unclear. In this study, we investigated the mechanistic role of purinergic
Minh T Do et al.
Environmental health perspectives, 113(4), 418-424 (2005-04-07)
Chlorination disinfection by-products (CDBPs) are produced during the treatment of water with chlorine to remove bacterial contamination. CDBPs have been associated with an increased risk of bladder cancer. There is also some evidence that they may increase the risk of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service