20338
Porapak™ Porous Polymer Adsorbent
matrix Porapak S, 100-120 mesh, bottle of 75 cc
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
Quality Level
packaging
bottle of 75 cc
parameter
250 °C temp. limit
technique(s)
gas chromatography (GC): suitable
surface area
~375 m2/g
matrix
Porapak S
particle size
100-120 mesh
density
~0.35 g/mL (free fall density)
Looking for similar products? Visit Product Comparison Guide
General description
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. Porapak™ S is a porous polymer composed of vinyl pyridine and is most suitable for separation of normal and branched alcohols.
Porapak porous polymers are spherical beads with consistent particle size, porosity, and surface area that help to insure analytical precision and accuracy.
For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
Application
Porapak™ S maybe used in isothermal gas chromatographic analysis of light gases, for detection and separation of CO2, C2H2, C2H4 and C2H6.
Legal Information
PoraPak is a trademark of Waters
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gas Chromatography, 127--127- (2012)
A new column system for isothermal gas chromatographic analysis of light gases (H2, O2, N2, CO, CH4, CO2, C2H4, C2H6 and C2H2) employing a column switch technique.
Chromatographia, 4 (7), 279-285 (1971)
Modern Practice of Gas Chromatography, 79-83 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service