Skip to Content
Merck
All Photos(1)

Documents

20301

Supelco

Molecular Sieve, 5Å

45-60 mesh, bottle of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
eCl@ss:
32110204

product name

Molecular Sieve Adsorbent, matrix Molecular Sieve 5A, 45-60 mesh, bottle of 50 g

Agency

suitable for USP/NF nitrogen

Quality Level

form

solid

packaging

bottle of 50 g

matrix

Molecular Sieve 5A

particle size

45-60 mesh

pore size

5 Å pore diameter

density

~0.72 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Molecular sieves are synthetically produced zeolites (naturally occurring aluminosilicate minerals), and are characterized by pores and internal cavities of extremely uniform dimensions. These crystalline materials have three-dimensional structures based on silicon oxide (SiO4) and aluminum oxide (AlO4) polyhedra. The polyhedra are linked by their corners to produce an open structure with internal cavities in which molecules can be trapped. These materials are engineered so that access to the internal cavities is through specific and uniform sized pores.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of atmospheric nitrous oxide using a molecular sieve 5A trap and gas chromatography.
Miles LD.
Analytical Chemistry, 43 (8), 1113-1115 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service