Skip to Content
Merck
All Photos(1)

Documents

20298

Supelco

Molecular Sieve, 5Å

water vapor trap refill, volume ~460 cc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
eCl@ss:
32110204

product name

Molecular Sieve 5A Water Vapor Trap Refill, volume ~460 cc

form

solid

Quality Level

technique(s)

gas chromatography (GC): suitable

volume

~460 cc

particle size

8-12 mesh

pore size

5 Å

General description

Mesh size is 8-12 mesh.
Molecular Sieve 5A.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service