00032
1H-1,2,4-Triazole-13C2,15N3
PESTANAL®, analytical standard
Synonym(s):
1H-1,2,4-Triazole-1,2,4-15N3-3,5-13C2
About This Item
Recommended Products
grade
analytical standard
product line
PESTANAL®
Assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
storage temp.
2-8°C
SMILES string
[H][15N]1[15N]=[13C]([H])[15N]=[13C]1[H]
[H][15N]1[15N]=[13C]([H])[15N]=[13C]1[H]
InChI
1S/C2H3N3/c1-3-2-5-4-1/h1-2H,(H,3,4,5)/i1+1,2+1,3+1,4+1,5+1
InChI key
NSPMIYGKQJPBQR-CVMUNTFWSA-N
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service