Skip to Content
Merck

Skip To

1.00264

Formic acid

greener alternative

98%-100%, for analysis EMSURE ACS,Reag. Ph Eur

Synonym(s):

Formic acid, Methanoic acid, Formylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
UNSPSC Code:
12352106
EC Index Number:
200-579-1
NACRES:
NA.21
Beilstein/REAXYS Number:
1209246
MDL number:
Assay:
≥98% (alkalimetric)
Form:
liquid
Grade:
ACS reagent
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Formic acid 98-100%, for analysis EMSURE® ACS,Reag. Ph Eur

grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

product line

EMSURE®

assay

≥98% (alkalimetric)

form

liquid

autoignition temp.

1004 °F

expl. lim.

12-38 % (v/v), 57 %

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable

impurities

≤500 ppm Acetic acid (CH3COOH)

evapn. residue

≤10 ppm

color

APHA: ≤10

refractive index

n20/D 1.370 (lit.)

pH

2.2 (20 °C, 10 g/L in H2O)

bp

100-101 °C (lit.), 101 °C/1013 hPa

mp

8.2-8.4 °C (lit.)

transition temp

flash point 49.5 °C

solubility

water: soluble

density

1.22 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm, sulfate (SO42-): ≤5 ppm, sulfite (SO32-): ≤10 ppm

SMILES string

OC=O

cation traces

Ag: ≤0.020 ppm, Al: ≤0.050 ppm, Ba: ≤0.050 ppm, Be: ≤0.020 ppm, Bi: ≤0.10 ppm, Ca: ≤0.20 ppm, Cd: ≤0.050 ppm, Co: ≤0.020 ppm, Cr: ≤0.050 ppm, Cu: ≤0.020 ppm, Fe: ≤2.0 ppm, Ge: ≤0.050 ppm, K: ≤0.10 ppm, Li: ≤0.020 ppm, Mg: ≤0.50 ppm, Mn: ≤0.050 ppm, Mo: ≤0.020 ppm, NH4+: ≤10 ppm, Na: ≤0.50 ppm, Ni: ≤0.050 ppm, Pb: ≤0.020 ppm, Sr: ≤0.020 ppm, Ti: ≤0.10 ppm, Tl: ≤0.050 ppm, V: ≤0.050 ppm, Zn: ≤0.050 ppm, Zr: ≤0.10 ppm, heavy metals (as Pb): ≤10 ppm

greener alternative category

storage temp.

15-25°C

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.
for analysis EMSURE® ACS,Reag. Ph Eur

Application


  • Modified QuEChERS method combined with ultra performance liquid chromatography-tandem mass spectrometry for detection of cyclopiazonic acid in feeds: This study demonstrates the use of 98-100% formic acid in a modified QuEChERS method, enhancing the efficiency of toxin detection in animal feeds, crucial for maintaining food safety and environmental health (Peng et al., 2024).

  • Adapting Fabric Phase Sorptive Extraction as an Innovative Multitool for Sample Transfer and Extraction in Pharmacokinetic Analysis Followed by LC-MS Determination of Levofloxacin in Plasma Samples: This study utilizes formic acid in developing advanced extraction methods for pharmacokinetic analysis, critical for drug development and therapeutic monitoring in pharmaceutical research (Ekin Dolaksız et al., 2024).

  • Stability Indicating Method Development and Validation of Glycyrrhizin Using RP-HPLC-DAD: Application to Glycyrrhiza glabra Extract: This research uses formic acid in chromatographic analyses to ensure the stability and quality of herbal medicine compounds, relevant in both pharmaceutical and biotechnological fields (Lyngdoh et al., 2024).

Analysis Note

Assay (alkalimetric)≥ 98.0 %Color≤ 10 HazenAcetic acid (CH₃COOH)≤ 500 ppmChloride (Cl)≤ 5 ppmSulfate (SO₄)≤ 5 ppmSulfite (SO₃)≤ 10 ppmHeavy metals (as Pb)≤ 10 ppmAg (Silver)≤ 0.020 ppmAl (Aluminium)≤ 0.050 ppmBa (Barium)≤ 0.050 ppmBe (Beryllium)≤ 0.020 ppmBi (Bismuth)≤ 0.10 ppmCa (Calcium)≤ 0.20 ppmCd (Cadmium)≤ 0.050 ppmCo (Cobalt)≤ 0.020 ppmCr (Chromium)≤ 0.050 ppmCu (Copper)≤ 0.020 ppmFe (Iron)≤ 2.0 ppmGe (Germanium)≤ 0.050 ppmK (Potassium)≤ 0.10 ppmLi (Lithium)≤ 0.020 ppmMg (Magnesium)≤ 0.50 ppmMn (Manganese)≤ 0.050 ppmMo (Molybdenum)≤ 0.020 ppmNH₄ (Ammonium)≤ 10 ppmNa (Sodium)≤ 0.50 ppmNi (Nickel)≤ 0.050 ppmPb (Lead)≤ 0.020 ppmSr (Strontium)≤ 0.020 ppmTi (Titanium)≤ 0.10 ppmTl (Thallium)≤ 0.050 ppmV (Vanadium)≤ 0.050 ppmZn (Zinc)≤ 0.050 ppmZr (Zirconium)≤ 0.10 ppmEvaporation residue≤ 10 ppmDilution testpasses testACS, Ph Eur-Reagent
Expiry date : see product label

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.43804Y0001970PHR3559
grade

ACS reagent

grade

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

assay

≥98% (alkalimetric)

assay

≥98% (alkalimetric)

assay

-

assay

-

Quality Level

300

Quality Level

100

Quality Level

-

Quality Level

300

agency

reag. Ph. Eur.

agency

suitable for DIN 38407-42, suitable for ISO/CEN 15968-2010

agency

-

agency

traceable to EP Y0001970, traceable to USP 1283200

form

liquid

form

liquid

form

-

form

-

pH

2.2 (20 °C, 10 g/L in H2O)

pH

2.2 (20 °C, 10 g/L in H2O)

pH

-

pH

-


Still not finding the right product?

Explore all of our products under Formic acid 98-100%


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

121.1 °F - closed cup

flash_point_c

49.5 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters