Skip to Content
Merck
All Photos(8)

Key Documents

WH0023435M1

Sigma-Aldrich

Monoclonal Anti-TARDBP antibody produced in mouse

clone 2E2-D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TAR DNA binding protein, Anti-TDP43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E2-D3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARDBP(23435)

General description

HIV-1, the causative agent of acquired immunodeficiency syndrome (AIDS), contains an RNA genome that produces a chromosomally integrated DNA during the replicative cycle. Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element (TAR) located downstream of the transcription initiation site. The protein encoded by this gene is a transcriptional repressor that binds to chromosomally integrated TAR DNA and represses HIV-1 transcription. In addition, this protein regulates alternate splicing of the CFTR gene. A similar pseudogene is present on chromosome 20. (provided by RefSeq)
TAR DNA binding protein 43 (TDP-43) belongs to the heterogeneous nuclear ribonucleoprotein family. It is expressed in neurons, glia and other cell types. This gene is located on human chromosome 1p36.

Immunogen

TARDBP (NP_031401.1, 1 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSEYIRVTEDENDEPIEIPSEDDGTVLLSTVTAQFPGACGLRYRNPVSQCMRGVRLVEGILHAPDAGWGNLVYVVNYPKDNKRKMDETDASSAVKVKRAVQKTSDLIVLGLPWKTTEQDLKEYFSTFGEVLMVQVKKDLKTGHSKGFGFVRFTEYETQVKVMSQRHMIDGRWCDCKLPNSKQSQDEPLRSRKVFVGRCTEDMTEDELREFFSQYGDVMDVFIPKPFRAFAFVTFADDQIAQSLCGEDLIIKGISVHISNA

Application

Monoclonal Anti-TARDBP antibody has been used in immunohistochemistry and immunoblotting.

Biochem/physiol Actions

TAR DNA binding protein 43 (TDP-43) participates in RNA alternative splicing, stability, transcriptional regulation, mRNA transport and translation. It is associated with Alzheimer′s disease. Mutations in TDP-43 result in proteinaceous inclusions in neurons and are expected to cause frontotemporal dementia or amyotrophic lateral sclerosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rates of hippocampal atrophy and presence of post-mortem TDP-43 in patients with Alzheimer's disease: a longitudinal retrospective study
Josephs KA, et al.
Lancet Neurology, 16(11), 917-924 (2017)
Mutations in UBQLN2 cause dominant X-linked juvenile and adult-onset ALS and ALS/dementia
Deng HX, et al.
Nature, 477(7363), 211-215 (2011)
Drosophila lines with mutant and wild type human TDP-43 replacing the endogenous gene reveals phosphorylation and ubiquitination in mutant lines in the absence of viability or lifespan defects
Chang JC and Morton DB
PLoS ONE, 12(7) (2017)
Tauana Bernardes Leoni et al.
Annals of neurology, 90(2), 239-252 (2021-05-29)
Protein misfolding plays a central role not only in amyotrophic lateral sclerosis (ALS), but also in other conditions, such as frontotemporal dementia (FTD), inclusion body myopathy (hIBM) or Paget's disease of bone. The concept of multisystem proteinopathies (MSP) was created
CSPα promotes SNARE-complex assembly by chaperoning SNAP-25 during synaptic activity
Sharma M, et al.
Nature Cell Biology, 13(1), 30-39 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service