Skip to Content
Merck
All Photos(3)

Key Documents

T1705

Sigma-Aldrich

Anti-TDP-43 antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ALS10, Anti-TARDBP, Anti-TARDP43, Anti-Tar DNA binding protein 43

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human, rat, mouse

concentration

~1.5 mg/mL

technique(s)

immunohistochemistry: 5-10 μg/mL using rat, mouse or human kidney
indirect immunofluorescence: 5-10 μg/mL using human HepG2 cells
western blot: 1.5-3.0 μg/mL using human U2OS cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TARDBP(23435)

Related Categories

General description

TDP-43 (TAR DNA binding protein, TARDP) is encoded by the gene mapped to human chromosome 1p36.22. The encoded protein belongs to the family of heterogenous nuclear ribonucleoproteins (hnRNPs) that bind single stranded RNA. TDP-43 is a 414 amino acid nuclear protein and is widely expressed in a variety of tissues.

Specificity

Anti-TDP-43 specifically recognizes human, mouse, and rat TDP-43.

Biochem/physiol Actions

Heterogeneous nuclear ribonucleoproteins (hnRNPs) are involved in the generation and processing of RNA, including transcription, splicing, transport and stability. TDP-43 regulates transcription of human immunodeficiency virus (HIV). The protein is identified as a major pathological protein, in both frontotemporal lobe degeneration subtype (FTLD-U) and amyotrophic lateral sclerosis (ALS). Abnormal phosphorylation of TDP-43 at Ser409/410 has also been observed in FTLD-U and ALS, suggesting a toxic gain of function leading to apoptosis.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J G Eldridge et al.
PLoS pathogens, 17(12), e1010173-e1010173 (2021-12-21)
For many intracellular bacterial pathogens manipulating host cell survival is essential for maintaining their replicative niche, and is a common strategy used to promote infection. The bacterial pathogen Listeria monocytogenes is well known to hijack host machinery for its own
Miao Sun et al.
Journal of neuroscience research, 92(1), 54-63 (2013-11-23)
The 43-kDa transactivation response DNA binding protein (TDP43), fused in sarcoma/translocated in liposarcoma (FUS/TLS), heat shock protein 70 (HSP70), and β-amyloid (Aβ) are induced and involved in cerebral ischemia, amyotrophic lateral sclerosis (ALS), and Alzheimer's disease (AD), but their relationships
TDP-43 regulates retinoblastoma protein phosphorylation through the repression of cyclin-dependent kinase 6 expression
Ayala YM, et al.
Proceedings of the National Academy of Sciences, 105(10), 3785-3789 (2008)
Rodger Wilhite et al.
Future science OA, 3(4), FSO238-FSO238 (2017-11-15)
Alzheimer's disease (AD) and other forms of dementia create a noncurable disease population in world's societies. To develop a blood-based biomarker is important so that the remedial or disease-altering therapeutic intervention for AD patients would be available at the early
The role of TDP-43 in amyotrophic lateral sclerosis and frontotemporal dementia
Mackenzie, Ian RA and Rademakers, Rosa
Current Opinion in Neurology, 21(6), 693-693 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service