Skip to Content
Merck
All Photos(7)

Key Documents

T6557

Sigma-Aldrich

Anti-γ-Tubulin Antibody

mouse monoclonal, GTU-88

Synonym(s):

Anti-γ-Tubulin antibody, Mouse monoclonal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

GTU-88, monoclonal

mol wt

antigen 48 kDa

contains

15 mM sodium azide

species reactivity

rat, hamster, chicken, human, bovine, canine, Xenopus, mouse

technique(s)

immunocytochemistry: 1:5,000-1:10,000 using HeLa cells
indirect ELISA: suitable
western blot: 1:10,000 using cultured chicken fibroblast extract

isotype

IgG1

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBG1(7283)
mouse ... Tubg1(103733)
rat ... Tubg1(252921)

Looking for similar products? Visit Product Comparison Guide

General description

γ-Tubulin (48kDa) is a ubiquitous and highly conserved protein within the microtubule organizing centers (MTOCs) in eukaryotic cells. γ-Tubulin is mapped to human chromosome 17q21.2 and codes for a member of the tubulin family. Human TUBG1 transcript is widely expressed in preimplantation embryos and brain.
Monoclonal Anti-γ-Tubulin (mouse IgG1 isotype) is derived from the GTU-88 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Specificity

Monoclonal Anti-γ-Tubulin recognizes an epitope located in the N-terminal amino acids of γ-tubulin (48 kDa). Cross reactivity has been observed with human, bovine, dog, hamster, rat, mouse, chicken, and Xenopus γ-tubulin.
The antibody recognizes an epitope located within the N-terminal region of γ-tubulin.

Immunogen

synthetic γ-tubulin peptide, conjugated to KLH

Application

Monoclonal Anti-γ-Tubulin antibody has been used in western blotting, indirect immunofluorescence (IF) and immunofluorescence staining.
Monoclonal Anti-γ-Tubulin is also suitable for use in immunochemical applications such as immunoblotting, immunocytochemical staining of cultured cells and in ELISA.
Monoclonal Anti-gamma-Tubulin is suitable for use in immunochemical applications such as immunoblotting, immunocytochemical staining of cultured cells, and in ELISA.

Biochem/physiol Actions

γ-Tubulin nucleates microtubule assembly throughout the mammalian cell cycle in vivo. γ-Tubulin binds microtubule minus ends and is responsible for mediating the link between microtubules and the centrosome. It functions as the microtubule nucleator at the MTOC. It binds to the β-tubulin half of the tubulin molecule, thus establishing the polarity of a microtubule, leaving the α-tubulin half exposed at the plus end. γ-Tubulin abundance is less than 1% of the level of either α- or β-tubulin. Overexpression of γ-tubulin is observed in lung cancer. The expression levels of γ-tubulin can be considered as an important prognostic indicator for patients with astrocytomas.

Physical form

The product is provided as ascites fluid with 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in "frostfree" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of γ-tubulin in non-small cell lung cancer.
Maounis NF, et al.
Histology and Histopathology, 27(7), 1183-1183 (2012)
Human mineralocorticoid receptor (MR) gene haplotypes modulate MR expression and transactivation: implication for the stress response.
Van Leeuwen N, et al.
Psychoneuroendocrinology, 36(5), 699-709 (2011)
Downregulation of connexin43 by microRNA-130a in cardiomyocytes results in cardiac arrhythmias
Osbourne A, et al.
Journal of Molecular and Cellular Cardiology, 74, 53-63 (2014)
Gamma-tubulin is a minus end-specific microtubule binding protein.
Li Q and Joshi HC
The Journal of Cell Biology, 131(1), 207-214 (1995)
γ-Tubulin is a component of the spindle pole body that is essential for microtubule function in Aspergillus nidulans.
Oakley BR, et al.
Cell, 61(7), 1289-1301 (1990)

Articles

Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α-and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service