Skip to Content
Merck
All Photos(7)

Key Documents

T6199

Sigma-Aldrich

Anti-TUBA4A (TUBA1) Antibody

enhanced validation

mouse monoclonal, DM1A

Synonym(s):

Monoclonal Anti-α-Tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DM1A, monoclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

yeast, mouse, amphibian, human, rat, chicken, fungi, bovine

packaging

antibody small pack of 25 μL

enhanced validation

independent ( Antibodies)
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

immunocytochemistry: 0.5-1 μg/mL using cultured chicken fibroblasts (CFB)
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 0.5-1 μg/mL using total tissue extract from chicken gizzard

isotype

IgG1

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBA4A(7277)
mouse ... Tuba1a(22142)
rat ... Tuba1a(64158)

Looking for similar products? Visit Product Comparison Guide

General description

Anti-α-Tubulin antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma DM1A produced by the fusion of mouse myeloma cells (NS1) and splenocytes from BALB/c mice immunized with purified chick brain tubulin. The isotype is determined by a double diffusion immunoassay using Mouse Monoclonal Antibody Isotyping Reagents, Product Number ISO2.

Tubulin is the major building block of microtubules. This intracellular, cylindrical, filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and the cytoskeleton. Tubulin is a heterodimer that consists of α-tubulin and β-tubulin. Both subunits have a molecular weight of approx. 50 kDa and share considerable homology. In addition to α- and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven. Most of these tubulins have distinct subcellular localization and an emerging diverse set of functions. Out of the seven different tubulins four new members of the tubulin family were identified recently, which consist of δ, ξ, η and ε-tubulin. η and ε-tubulins were discovered by database searches. Microtubular systems contain at least three α-tubulin isoforms. Two isoforms are coded by two α-tubulin genes, which are both transcribed and code for extremely similar proteins. The third isoform is generated by post-translational modification. At least three modifications of tubulin subunits have been described: the phosphorylation of β-tubulin from brain, the removal of the carboxy terminal tyrosine form a-tubulin in vertebrate tissues, and the acetylation of the amino group of lysine(s) in α-tubulin.

α -tubulin, also called tubulin α 4a (TUBA4A), is mapped to human chromosome 2q35. The gene codes for a member of the α -tubulin family, and contains 448 amino acids. α-subunit of tubulin has molecular weight of 50,000.

Monoclonal antibodies recognizing α-tubulin, together with monoclonal antibodies to other tubulin types (β, β-tubulin isotype I +II, β-tubulin isotype III, tyrosine tubulin, and the acetylated form of α-tubulin) provide a specific and useful tool in studying the intracellular distribution of tubulin and the static and dynamic aspects of cytoskeleton.

Specificity

Anti-α-Tubulin antibody, Mouse monoclonal recognizes an epitope located at the C-terminal end of the α-tubulin isoform (amino acids 426-430) in a variety of organisms (e.g., human, bovine, mouse, and chicken).
The antibody is specific for α-tubulin in immunoblotting assays and may be used for localization of α-tubulin in cultured cells or tissue sections. The antibody reacts best with chicken fibroblasts.

Immunogen

microtubules from chicken embryo brain.

Application

Anti-α-Tubulin antibody, Mouse monoclonal has been used in immunocytofluorescence and western blot analysis.

Biochem/physiol Actions

Tubulin α 4a (TUBA4A) mutation is associated with amyotrophic lateral sclerosis (ALS).

α-Tubulin or tubulin α 4A (TUBA4A) plays an essential role in cell growth and division. It is also implicated in various cellular processes, such as cell motility, signaling development and cell shape maintenance. Mutation in TUBA4A is associated with the development of various types of cancers, such as oral cancer, breast cancer, rectal cancer, lung cancer and prostate cancer. In addition, variation in the TUBA4A leads to sporadic amyotrophic lateral sclerosis (ALS).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Preparation Note

Storage and Stability: Store at 2-8°C for up to one month. For prolonged storage, freeze in working aliquots at -20°C. Repeated freezing and thawing, or storage in frost-free freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Other Notes

In order to obtain the best results using various techniques and preparations, we recommend determining optimal working dilutions by titration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise K Reaves et al.
PloS one, 9(3), e91747-e91747 (2014-03-19)
The claudin-low molecular subtype of breast cancer is of particular interest for clinically the majority of these tumors are poor prognosis, triple negative, invasive ductal carcinomas. Claudin-low tumors are characterized by cancer stem cell-like features and low expression of cell
Kun Yang et al.
Schizophrenia research, 210, 149-156 (2019-06-18)
DISC1 was originally expected to be a genetic risk factor for schizophrenia, but the genome wide association studies have not supported this idea. In contrast, neurobiological studies of DISC1 in cell and animal models have demonstrated that direct perturbation of
Exercise alters SIRT1, SIRT6, NAD and NAMPT levels in skeletal muscle of aged rats.
Koltai E, et al.
Mechanisms of Ageing and Development, 131(1), 21-28 (2010)
A role for spleen monocytes in post-ischemic brain inflammation and injury.
Bao Y, et al.
Journal of Neuroinflammation, 7(1), 92-92 (2010)
Stéphane G Rolland et al.
Cell reports, 28(7), 1659-1669 (2019-08-15)
The induction of the mitochondrial unfolded protein response (UPRmt) results in increased transcription of the gene encoding the mitochondrial chaperone HSP70. We systematically screened the C. elegans genome and identified 171 genes that, when knocked down, induce the expression of an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service