Skip to Content
Merck
All Photos(1)

Key Documents

T2255

Sigma-Aldrich

Trifluorothymidine

≥99% (HPLC)

Synonym(s):

α,α,α-Trifluorothymidine, 2′-Deoxy-5-trifluoromethyluridine, Trifluorothymine deoxyriboside, Trifluridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11F3N2O5
CAS Number:
Molecular Weight:
296.20
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Assay

≥99% (HPLC)

form

powder

mp

190-193 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C(=O)NC2=O)C(F)(F)F

InChI

1S/C10H11F3N2O5/c11-10(12,13)4-2-15(9(19)14-8(4)18)7-1-5(17)6(3-16)20-7/h2,5-7,16-17H,1,3H2,(H,14,18,19)/t5-,6+,7+/m0/s1

InChI key

VSQQQLOSPVPRAZ-RRKCRQDMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trifluorothymidine has been used as a component of selection medium to screen thymidine kinase (TK) mutants in lymphoma cells, human lymphoblastoid TK6 cells, and L5178Y mouse lymphoma cells.

Biochem/physiol Actions

Trifluorothymidine is a thymidine analog and is light sensitive. TFT serves as a thymidine kinase substrate to study enzyme specificity and kinetics. Incorporation of phosphorylated TFT into DNA induces damage, making it useful for DNA repair studies. TFT may also be used in the inhibition of thymidylate synthase and in screening mutant thymidine kinase gene.. It elicits antitumor activity in gastrointestinal (GI) cancers and has therapeutic potential to treat herpetic keratitis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Carmine et al.
Drugs, 23(5), 329-353 (1982-05-01)
Trifluridine (trifluorothymidine) is an antiviral agent for topical use in the eye, and is structurally related to idoxuridine. In vitro studies have shown that it effectively inhibits the replication of herpes simplex virus type 1, which causes primary keratoconjunctivitis and
Olaf H Temmink et al.
Molecular cancer therapeutics, 9(4), 1047-1057 (2010-04-08)
Trifluorothymidine (TFT) is part of the novel oral formulation TAS-102, which is currently evaluated in phase II studies. Drug resistance is an important limitation of cancer therapy. The aim of the present study was to induce resistance to TFT in
Marc Peeters et al.
Future oncology (London, England), 14(16), 1629-1645 (2018-04-28)
Fluoropyrimidines are currently the backbone of treatment for gastrointestinal (GI) cancers but development of resistance to these agents remains a major problem. Trifluridine/tipiracil is an oral chemotherapeutic agent recently approved for third-line treatment of chemorefractory metastatic colorectal cancer. This article
I V Bijnsdorp et al.
Nucleosides, nucleotides & nucleic acids, 27(6), 699-703 (2008-07-05)
Trifluorothymidine (TFT), a potent anticancer agent, inhibits thymidylate synthase (TS) and is incorporated into the DNA, both events resulting in cell death. Cell death induction related to DNA damage often involves activation of p53. We determined the role of p53
Connie Kang et al.
Drugs, 79(14), 1583-1590 (2019-09-07)
Trifluridine/tipiracil (Lonsurf®) is a fixed-dose combination tablet comprising trifluridine, an antineoplastic nucleoside analogue, and tipiracil, a thymidine phosphorylase inhibitor. Trifluridine/tipiracil has recently been granted an additional indication in the USA for the treatment of metastatic gastric cancer, including gastroesophageal junction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service