Skip to Content
Merck
All Photos(1)

Key Documents

S2067

Sigma-Aldrich

S9 from Liver, Pooled

from rat(Sprague-Dawley), male

Synonym(s):

Liver S9 Fraction, S9 Liver Extract

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.47

biological source

rat (Sprague-Dawley)

Quality Level

packaging

vial of ~20 mg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

S9 from Liver, Pooled has been used in metabolic activation in Salmonella typhimurium strains.

Biochem/physiol Actions

The hepatic S9 pools from a variety of biological sources represent the post-mitochondrial supernatant fraction from homogenized liver. Known to be a rich source of drug metabolizing enzymes including P-450, these pools are useful in the study of xenobiotic metabolism and drug interactions.
They are used in the Ames test, which is a method that uses bacteria to test if a given chemical causes mutations in the genetic material of a given test organism.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genotoxicity evaluation of locally produced dental porcelain-An in vitro study using the Ames and Comet assays
Noushad M, et al.
Toxicology in vitro, 23(6), 1145-1150 (2009)
The Ames Salmonella/microsome mutagenicity assay.
Mortelman K and Zeiger E
Mutation Research, 455(1-2), 29-60 (2000)
Esra Erikel et al.
Environmental and molecular mutagenesis, 64(5), 291-308 (2023-05-10)
Amygdalin (AMY), a plant secondary metabolite containing nitrile, is a major component of the seeds of Rosaceae family plants. It is known that this compound has many pharmacological activities such as cancer prevention, antipyretic, and cough suppressant. In this study
Leonara Fayer et al.
Toxicology research, 10(3), 511-522 (2021-06-19)
The titanium dioxide nanoparticles (NPs) have been applied to biomedical, pharmaceutical, and food additive fields. However, the effect on health and the environment are conflicting; thus, it has been reviewing several times. In this context, establishing standard robust protocols for
Milena Lopes Francisco Bittencourt et al.
Life sciences, 256, 117977-117977 (2020-07-01)
Silibinin is the major component of flavonolignans complex mixture (Silymarin), which is obtained from Silybum marianum (L.) Gaertn. Despite several reports about silibinin, little is known about its effects on gastric diseases. Then, the present study aims to evaluate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service