Skip to Content
Merck
All Photos(4)

Key Documents

A1888

Sigma-Aldrich

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt

≥98% (HPLC), chromogenic, powder

Synonym(s):

AzBTS-(NH4)2, Diammonium 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N6O6S4
CAS Number:
Molecular Weight:
548.68
Beilstein:
7329461
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

solubility

water: 10 mg/mL, clear to slightly hazy

storage temp.

2-8°C

SMILES string

[NH4+].[NH4+].CCN1C(\Sc2cc(ccc12)S([O-])(=O)=O)=N\N=C3/Sc4cc(ccc4N3CC)S([O-])(=O)=O

InChI

1S/C18H18N4O6S4.2H3N/c1-3-21-13-7-5-11(31(23,24)25)9-15(13)29-17(21)19-20-18-22(4-2)14-8-6-12(32(26,27)28)10-16(14)30-18;;/h5-10H,3-4H2,1-2H3,(H,23,24,25)(H,26,27,28);2*1H3/b19-17-,20-18-;;

InChI key

OHDRQQURAXLVGJ-AXMZSLBLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ABTS, 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid), is water soluble chemical compound used as chromogenic substrate:
  • ABTS together with hydrogen peroxide for reaction with peroxidase enzymes (like HRP for example).
  • ABTS together with laccase or bilirubin oxidase.

ABTS mainly used in ELISA (enzyme-linked immunosorbent assay) procedures.
A peroxidase reaction of ABTS in the presence of hydrogen peroxide produces a green soluble end product which can be read spectrophotometrically at 405nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS).††† ABTS, 2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt is light sensitive.

Application

2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is green in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 1% sodium dodecyl sulfate (SDS). Recommended for ELISA (microwell) procedures, not recommended for membrane applications.
  • 2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt has been used as a substrate in ELISA (enzyme-linked immunosorbent assay).
  • ABTS is significantly used by the food industry, to measure the antioxidant capacities within different foods.
  • ABTS is also used to monitor glucose concentrations in different solutions including blood serum.
  • It is recommended for ELISA (microwell) procedures, not recommended for membrane applications.Since it is less readily oxidized than TMB and OPD substrates, ABTS is less sensitive for ELISA applications. Thus ABTS has some advantageous over TMB and OPD in cases of large background results.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of glucocorticoid resistance following social stress requires a second signal.
Avitsur R et al.
Journal of Leukocyte Biology, 74, 507-507 (2003)
Fan Shi et al.
Biotechnology letters, 42(5), 825-832 (2020-01-30)
To effectively and conveniently detect pathogenic bacteria, this study aimed to develop label-free biosensors fabricated affinity peptides that can recognize targeted bacteria strains and enable precise quantitative detections. A 12-mer peptide with high binding affinity toward Escherichia coli O157:H7 was
Structure-based mutagenesis of herpes simplex virus glycoprotein D defines three critical regions at the gD-HveA/HVEM binding interface.
Connolly SA et al.
Journal of Virology, 77, 8127-8127 (2003)
Evaluation of ELISA with ABTS, 2-2'-azino-di-(3-ethylbenzthiazoline sulfonic acid), as the substrate of peroxidase and its application to the diagnosis of schistosomiasis.
Matsuda H et al.
The Japanese Journal of Experimental Medicine, 54, 131-131 (1984)
Crowther JR
ELISA: Theory and Practice (1995)

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service