Skip to Content
Merck
All Photos(1)

Key Documents

SML3605

Sigma-Aldrich

Selexipag

≥98% (HPLC)

Synonym(s):

2-{4-[(5,6-Diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}-N-(methylsulfonyl)acetamide, ACT-293987, NS-304

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N4O4S
CAS Number:
Molecular Weight:
496.62
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

CC(C)N(CCCCOCC(NS(C)(=O)=O)=O)C1=NC(C2=CC=CC=C2)=C(C3=CC=CC=C3)N=C1

Biochem/physiol Actions

Selexipag is a potent, highly selective and orally available nonprostanoid agonist for the PGI2 receptor (IP receptor). Selexipag is readily hydrolyzed to the active metabolite ACT-333679 (MRE-269), which is also a potent and selective agonist at IP receptors. In contrast to the prostacyclin analogs iloprost and beraprost, selexipag and metabolite ACT-333679 does not contract gastric fundus up to mM concentrations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A long-acting and highly selective prostacyclin receptor agonist prodrug, 2-{4-[(5,6-diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}-N-(methylsulfonyl)acetamide (NS-304), ameliorates rat pulmonary hypertension with unique relaxant responses of its active form, {4-[(5,6-diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}acetic acid (MRE-269), on rat pulmonary artery
Journal of Pharmacology and Experimental Therapeutics, 326(3), 691-699 (2008)
2-{4-[(5,6-Diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}-N-(methylsulfonyl)acetamide (NS-304), an Orally Available and Long-Acting Prostacyclin Receptor Agonist Prodrug
Journal of Pharmacology and Experimental Therapeutics, 322, 1181-1188 (2007)
Keith Morrison et al.
The Journal of pharmacology and experimental therapeutics, 335(1), 249-255 (2010-07-28)
Selexipag [2-{4-[(5,6-diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}-N-(methylsulfonyl)acetamide] is an orally available prostacyclin (PGI(2)) receptor (IP receptor) agonist that is chemically distinct from PGI(2) and is in clinical development for the treatment of pulmonary arterial hypertension. Selexipag is highly selective for the human IP receptor in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service