Skip to Content
Merck
All Photos(1)

Key Documents

R7388

Sigma-Aldrich

RPMI-1640 Medium

RPMI-1640 Medium

With HEPES, with ʟ-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

>7.2

components

HEPES: 20 mM
NaHCO3: no
sodium pyruvate: no
phenol red: yes
L-glutamine: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.
RPMI-1640 medium formulation is based on the RPMI-1630 series of media utilizing a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins. It is used for the culture of human normal and neoplastic leukocytes. RPMI-1640 also supports the growth of various types of cell cultures, like fresh human lymphocytes in the 72-hour phytohemagglutinin (PHA) stimulation assay.

Application

RPMI-1640 medium has been used to culture:
  • human immortalized keratinocyte cells and in in vitro transfection assay[1]
  • cell lines and human T-cell blasts and to monitor autophagic flux[2]
  • peripheral blood mononuclear cells of piglets (PBMCs)[3]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Linder et al.
PLoS genetics, 10(2), e1004207-e1004207 (2014-03-04)
RNA decay and maturation have in recent years been recognised as major regulatory mechanisms in bacteria. In contrast to Escherichia coli, the Firmicute (Gram-positive) bacteria often do not encode the well-studied endonuclease RNase E, but instead rely on the endonucleases
Janina Treffon et al.
Cellular microbiology, 22(5), e13158-e13158 (2020-01-03)
Staphylococcus aureus is one of the earliest pathogens that persists the airways of cystic fibrosis (CF) patients and contributes to increased inflammation and decreased lung function. In contrast to other staphylococci, S. aureus possesses two superoxide dismutases (SODs), SodA and
Maarit Suomalainen et al.
Journal of cell science, 134(5) (2020-09-13)
In clonal cultures, not all cells are equally susceptible to virus infection, and the mechanisms underlying this are poorly understood. Here, we developed image-based single-cell measurements to scrutinize the heterogeneity of adenovirus (AdV) infection. AdV delivers, transcribes and replicates a
Višnja Stepanić et al.
Antioxidants (Basel, Switzerland), 8(4) (2019-04-10)
Chalcones are polyphenolic secondary metabolites of plants, many of which have antioxidant activity. Herein, a set of 26 synthetic chalcone derivatives with alkyl substituted pyrazine heterocycle A and four types of the monophenolic ring B, were evaluated for the potential
Seda Tuncay Cagatay et al.
International journal of molecular sciences, 21(21) (2020-11-14)
Molecular communication between irradiated and unirradiated neighbouring cells initiates radiation-induced bystander effects (RIBE) and out-of-field (abscopal) effects which are both an example of the non-targeted effects (NTE) of ionising radiation (IR). Exosomes are small membrane vesicles of endosomal origin and

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service