Skip to Content
Merck
All Photos(1)

Key Documents

SML3476

Sigma-Aldrich

Brivaracetam

≥98% (HPLC)

Synonym(s):

(αS,4R)-α-Ethyl-2-oxo-4-propyl-1-pyrrolidineacetamide, (2S)-2-[(4R)-2-Oxo-4-propylpyrrolidin-1-yl]butanamide, (S)-2-((R)-2-Oxo-4-propylpyrrolidin-1-yl)butanamide, UCB 34714, UCB-34714, UCB34714

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O2
CAS Number:
Molecular Weight:
212.29
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule V

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Brivaracetam shows approximately 10 times greater potency than levetiracetam (Keppra)*; pIC50 at LBS = 7.0; inhibits audiogenic seizures in seizure-prone mice (ED50 = 14 μmol/kg) and is also effective in other animal models of epilepsy. Brivaracetam is used as an anticonvulsant (antiepileptic) for seizure treatment.
Orally active levetiracetam analog with higher synaptic vesicle protein 2A (SV2A) affinity and >10 times greater in vivo antiseizure efficay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of 4-substituted pyrrolidone butanamides as new agents with significant antiepileptic activity
Journal of medicinal chemistry, 47(3), 530-549 (2004)
Brivaracetam attenuates pain behaviors in a murine model of neuropathic pain
Molecular Pain (2019)
Brivaracetam is superior to levetiracetam in a rat model of post-hypoxic myoclonus
Journal of neural transmission (Vienna, Austria : 1996), 114(12), 1547-1551 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service