Skip to Content
Merck
All Photos(1)

Key Documents

SML0816

Sigma-Aldrich

Pramiracetam

≥98% (HPLC)

Synonym(s):

Amacetam, N-[2-(Diisopropylamino)ethyl]-2-(2-oxopyrrolidin-1-yl)acetamide, N-[2-[Bis(1-methylethyl)amino]ethyl]-2-oxo-1-pyrrolidineacetamide, Vinpotropil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H27N3O2
CAS Number:
Molecular Weight:
269.38
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])O.[N+H](C(C)C)(C(C)C)CCNC(=O)CN1CCCC1=O

InChI

1S/C14H27N3O2.H2O4S/c1-11(2)17(12(3)4)9-7-15-13(18)10-16-8-5-6-14(16)19;1-5(2,3)4/h11-12H,5-10H2,1-4H3,(H,15,18);(H2,1,2,3,4)

InChI key

ACSROKXFXFNERX-UHFFFAOYSA-N

Biochem/physiol Actions

Pramiracetam is a potent nootropic agent that is a member of the racetam drug family. Pramiracetam improves cognitive deficits associated with traumatic brain injuries. Also, pramiracetam is a specific inhibitor of prolyl endopeptidase.
Pramiracetam is a potent nootropic agent.
Pramiracetam plays an important role in spatial learning and memory in rats. It is considered as a memory enhancing agent and is stronger than piracetam.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pavlík et al.
Activitas nervosa superior, 29(1), 62-65 (1987-03-01)
High affinity choline uptake (HACU) in the hippocampus and striatal concentration of dopamine (DA) and homovanillic acid (HVA) as measures of the in vivo acetylcholine and DA turnover, respectively, were estimated in male rats, Long-Evans, following 6-day administration of various
C Mondadori et al.
Behavioural brain research, 34(1-2), 155-158 (1989-08-01)
The present experiments demonstrate that the absence of any memory-improving action of nootropics in adrenalectomized animals cannot be ascribed to an effect of dosage. Doses of 1, 10, 100, 1000 and 3000 mg/kg p.o. of piracetam, oxiracetam, aniracetam or pramiracetam
A Ennaceur et al.
Behavioural brain research, 33(2), 197-207 (1989-06-01)
The effects of the nootropic drugs Piracetam (Pir) and Pramiracetam (Pram) were evaluated on recognition-memory of rats in a new one-trial test. This test is based on spontaneous exploratory activity and does not involve rule learning or reinforcement. Recognition is
Z Fang et al.
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao, 30(4), 411-413 (2001-06-05)
Pharmacokinetic rules of pramiracetam were studied here. After giving pramiracetam orally to dogs, we drew their blood at various times. The drug concentrations in blood plasma were detected by HPLC. 3p87 program was used to calculate the pharmacokinetic parameters. The
B P Poschel et al.
Experientia, 41(11), 1433-1435 (1985-11-15)
The basal EEG profile of the aged Fisher-344 rat was consistently different from that of the young rat, showing dominant high voltage slow-wave components. These slow waves were present in both the frontal cerebral cortex and dorsal hippocampus. Absent or

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service