Skip to Content
Merck
All Photos(1)

Key Documents

SML3461

Sigma-Aldrich

Exatecan mesylate

≥98% (HPLC)

Synonym(s):

(1S,9S)-9-Ethyl-5-fluoro-9-hydroxy-4-methyl-10,13-dioxo-2,3,9,10,13,15-hexahydro-1H,12H-benzo[de]pyrano[3′,4′:6,7]indolizino[1,2-b]quinoline-1-amine methanesulfonate, DX8951f

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22FN3O4· CH3SO3H
CAS Number:
Molecular Weight:
531.55
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C1N2C(C3=NC4=CC(F)=C(C(CC[C@@H]5N)=C4C5=C3C2)C)=CC6=C1COC([C@@]6(CC)O)=O.OS(=O)(C)=O

InChI

1S/C24H22FN3O4.CH4O3S/c1-3-24(31)14-6-18-21-12(8-28(18)22(29)13(14)9-32-23(24)30)19-16(26)5-4-11-10(2)15(25)7-17(27-21)20(11)19;1-5(2,3)4/h6-7,16,31H,3-5,8-9,26H2,1-2H3;1H3,(H,2,3,4)/t16-,24-;/m0./s1

InChI key

BICYDYDJHSBMFS-GRGFAMGGSA-N

Biochem/physiol Actions

Exatecan mesylate is a water soluble camptothecin (CPT) analog that exhibits a stronger topoisomerase-I inhibitory and antitumor activities and is highly effective against P-glycoprotein (P-gp) mediated multi-drug resistant cells. Several of the exatecan derivative based antibody-drug and peptide-drug conjugate payloads display high therapeutic indices universally across solid tumors.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 1B - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Joto et al.
International journal of cancer, 72(4), 680-686 (1997-08-07)
We previously reported that DX-8951f, a novel water-soluble camptothecin analog, significantly inhibits the growth of various human and murine tumors in vitro and in vivo. The antitumor effects and topoisomerase I inhibitory activity of DX-8951f are stronger than those of
Jie Chen et al.
Molecular medicine reports, 19(1), 477-489 (2018-11-16)
Unresectable advanced pancreatic cancer (APC) is a highly lethal malignancy. Although numerous chemotherapeutic regimens are available, evidence regarding the survival extension, the life quality improvement, the associated risks and occurrence rates of adverse effects, is required. The effects of 19
Ghassan K Abou-Alfa et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 24(27), 4441-4447 (2006-09-20)
Exatecan mesylate is a hexacyclic, water-soluble, topoisomerase-1 inhibitor. Exatecan has single-agent and combination activity with gemcitabine in advanced pancreatic cancer. A multicenter, randomized, phase III trial comparing exatecan plus gemcitabine versus gemcitabine alone in advanced pancreatic cancer was conducted. Eligibility

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service