Skip to Content
Merck
All Photos(1)

Key Documents

SML3265

Sigma-Aldrich

DAPTA trifluoroacetate

≥95% (HPLC)

Synonym(s):

Adaptavir trifluoroacetate, D-Ala-Peptide T-NH2 trifluoroacetate, D-Ala-Peptide T-amide trifluoroacetate, Dala1-Peptide T-NH2 trifluoroacetate, Dala1-Peptide T-amide trifluoroacetate, H-D-Ala-STTTNYT-NH2 trifluoroacetate, H-D-Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr-NH2 trifluoroacetate, [D-Ala1]-Peptide T-NH2 trifluoroacetate, [D-Ala1]-Peptide T-amide trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

DAPTA is a potent CCR5 antagonist, an amidated octapeptide corresponding to an HIV envelope glycoprotein gp120 (gp160)-derived sequence (aa 185-192; the SF-2 isolate) that competes against gp120-CD4 complex for interaction with CCR5 (IC50 = 0.42 nM against gp120CM235 binding to CCR5/CD4 cells, IC50 = 1.5/1.8 nM against gp120BaL/sCD4 or gp120CM235/sCD4 binding to CCR5+/CD4- cells). DAPTA blocks CCR5-dependent HIV host cell entry in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiwei Yu et al.
Frontiers in pharmacology, 12, 551839-551839 (2021-05-07)
Background: Cigarette smoke exposure (CSE) is a major cause of chronic obstructive pulmonary disease (COPD). The smoke disrupts cell-cell adhesion by inducing epithelial barrier damage to the tight junction (TJ) proteins. Even though the inflammatory mechanism of chemokine (C-C motif)
Sheikh F Ahmad et al.
European journal of pharmacology, 846, 100-108 (2019-01-19)
Autism spectrum disorder (ASD) is a prevalent neurodevelopmental disorder characterized by deficits in social interaction, communication, and repetitive behaviors. BTBR T+ Itpr3tf/J (BTBR) mice, a preclinical autistic model featuring ASD symptoms as defined by social relations, was used in this
M R Ruff et al.
FEBS letters, 211(1), 17-22 (1987-01-19)
The octapeptide Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr (peptide T) and two structural analogs are potent agonists of human monocyte chemotaxis, evincing identical rank potency orders as was previously shown for their inhibition of human immunodeficiency virus (HIV) envelope binding and T cell infectivity. Chemotactic
C Liapi et al.
The Journal of investigative dermatology, 110(4), 332-337 (1998-04-16)
In addition to acquired immunodeficiency syndrome (AIDS), persons infected with human immunodeficiency virus often develop cutaneous manifestations, including severe psoriasis. In previous studies, we have established that psoriatic fibroblasts and erythrocytes obtained from psoriatic patients exhibit decreased levels of cyclic
Maria T Polianova et al.
Antiviral research, 67(2), 83-92 (2005-07-09)
The chemokine receptor CCR5 plays a crucial role in transmission of HIV isolates, which predominate in the early and middle stages of infection, as well as those, which populate the brain and cause neuro-AIDS. CCR5 is therefore an attractive therapeutic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service