Skip to Content
Merck
All Photos(1)

Key Documents

SML3261

Sigma-Aldrich

JS6

≥98% (HPLC)

Synonym(s):

2-(2-Fluorobenzamido)-N-[2-(morpholin-4-yl)ethyl]benzamide, 2-Fluoro-N-(2-(2-morpholinoethylcarbamoyl)phenyl)benzamide, 2-[(2-Fluorobenzoyl)amino]-N-[2-(4-morpholinyl)ethyl]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22FN3O3
CAS Number:
Molecular Weight:
371.41
UNSPSC Code:
12352107
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C1NC(C2=C(C=CC=C2)F)=O)NCCN3CCOCC3

InChI key

QEEPXSHOUBCRGQ-UHFFFAOYSA-N

Biochem/physiol Actions

JS6 is a Bcl3:NF-kB p50 protein-protein interaction (PPI) inhibitor that effectively suppresses Bcl3-dependent intracellular NF-kB activity (IC50 = 45/159 nM using Bcl3-overexpressing MDA-MB-231/HEK293; IC50 = 710 nM using HEK293 overexpressing Bcl3 and NF-kB p52). JS6 prevents enhanced MDA-MB-231 migration upon Bcl3 overexpression in cultures (IC50 = 310 nM), and effectively suppresses primary tumor growth as well as secondary tumor colonisation in vivo (3.5-10 mg/kg/d i.p. mice with MDA-MB-436 or 4T1.2 murine carcinoma xenograft) without overt systemic toxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jitka Soukupová et al.
Molecular cancer therapeutics, 20(5), 775-786 (2021-03-03)
The development of antimetastatic drugs is an urgent healthcare priority for patients with cancer, because metastasis is thought to account for around 90% of cancer deaths. Current antimetastatic treatment options are limited and often associated with poor long-term survival and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service