Skip to Content
MilliporeSigma

DPP4-M

DPP IV Inhibitor

Synonym(s):

DPP IV Inhibitor, Dipeptidyl peptidase 4 Inhibitor, Gliptins

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116012
NACRES:
NC.07
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

NCBI accession no.

UniProt accession no.

Gene Information

human ... DPP4(1803)

General description

Dipeptidyl peptidase IV (DPP IV), a dimeric type II integral membrane glycoprotein, is a member of the family of prolyl-specific proteases. It is abundantly expressed in the epithelial and nonepithelial tissues. It is highly expressed in the kidney and the colon.

Application

Research Category
All

Biochem/physiol Actions

Dipeptidyl peptidase IV (DPP IV) controls insulin-stimulating hormones, glucagon-like peptide (GLP-1), and glucose-dependent insulinotropic polypeptide (GIP). It′s a promising therapeutic target for type 2 diabetes (T2DM). In addition to its catalytic action, DPP-4 also serves as a binding protein and a ligand of extracellular factors. DPP-4 inhibition causes GLP-1 and GIP to have a prolonged activity, hence DPP-4 inhibitors help maintain glucose homeostasis. DPP IV inhibitors can improve glycemic control for a longer period when compared to early oral hypoglycemics.

Preparation Note

Upon arrival, store at 4°C. For long-term of more than 2 weeks, store at –20°C.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library