Skip to Content
Merck
All Photos(1)

Key Documents

SML3035

Sigma-Aldrich

Ziprasidone hydrochloride hydrate

≥97% (HPLC)

Synonym(s):

5-[2-[4-(1,2-Benzisothiazol-3-yl)-1-piperazinyl]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one hydrochloride hydrate, CP-88,059, CP-88,059-01, CP-88059

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClN4OS · xHCl · yH2O
CAS Number:
Molecular Weight:
412.94 (anhydrous free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

SMILES string

O=C1CC(C=C(C(Cl)=C2)CCN3CCN(CC3)C4=NSC5=C4C=CC=C5)=C2N1.[xHCl].[yH2O]

InChI

1S/C21H21ClN4OS/c22-17-13-18-15(12-20(27)23-18)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)28-24-21/h1-4,11,13H,5-10,12H2,(H,23,27)

InChI key

MVWVFYHBGMAFLY-UHFFFAOYSA-N

Biochem/physiol Actions

Ziprasidone is an atypical antipsychotic with 5HT1A agonist, as well as D2, 5HT2A, and 5HT1D antagonist activity.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Prakash et al.
Drug metabolism and disposition: the biological fate of chemicals, 25(2), 206-218 (1997-02-01)
The metabolism and excretion of ziprasidone (5-[2-[4-(1,2-benzisothiazol-3-yl)piperazin-1-yl]ethyl]-6-++ +chloroindolin-2-one hydrochloride hydrate) were studied in Long Evans rats after oral administration of a single dose of a mixture of 14C- and 3H-labeled ziprasidone. The radioactive dose was quantitatively recovered over 7 days
C J Bench et al.
Psychopharmacology, 124(1-2), 141-147 (1996-03-01)
Positron emission tomography (PET) and 11C-raclopride were used to assess the time course of binding to central dopamine D2 receptors by the novel neuroleptic ziprasidone. In a third party blind study, six healthy male control subjects received a predose of
T F Seeger et al.
The Journal of pharmacology and experimental therapeutics, 275(1), 101-113 (1995-10-01)
Ziprasidone (CP-88,059) is a combined 5-HT (serotonin) and dopamine receptor antagonist which exhibits potent effects in preclinical assays predictive of antipsychotic activity. Whereas the compound is a dopamine antagonist in vitro and in vivo, its most potent action is antagonism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service