Skip to Content
Merck
All Photos(1)

Key Documents

SML2904

Sigma-Aldrich

AGF347

≥98% (HPLC)

Synonym(s):

(4-(4-(2-Amino-4-oxo-3,4-dihydro-5H-pyrrolo[3,2-d]pyrimidin-5-yl)butyl)-2-fluorobenzoyl)-L-glutamic acid, (S)-2-(4-(4-(2-Amino-4-oxo-3H-pyrrolo[3,2-d]pyrimidin-5(4H)-yl)butyl)-2-fluorobenzamido)pentanedioic acid, N-[4-[4-(2-Amino-3,4-dihydro-4-oxo-5H-pyrrolo[3,2-d]pyrimidin-5-yl)butyl]-2-fluorobenzoyl]-L-glutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24FN5O6
CAS Number:
Molecular Weight:
473.45
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/L, clear

storage temp.

2-8°C

SMILES string

O=C(CC[C@H](NC(C1=CC=C(C=C1F)CCCCN2C=CC(N=C(N3)N)=C2C3=O)=O)C(O)=O)O

Biochem/physiol Actions

AGF347 is a potent inhibitor of serine hydroxymethyltransferase 2 (SHMT2) that inhibits mitochondrial C1 metabolism. It also inhibits cytosolic C1 targets such as SHMT1, GARFTase, and AICARFTase. AGF347 inhibits de novo purine biosynthesis. AGF347 exhibits potent broad spectrum in vitro and in vivo antitumor efficacy.
potent inhibitor of serine hydroxymethyltransferase 2 (SHMT2)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aamod S Dekhne et al.
Molecular cancer therapeutics, 18(10), 1787-1799 (2019-07-11)
Folate-dependent one-carbon (C1) metabolism is compartmentalized into the mitochondria and cytosol and supports cell growth through nucleotide and amino acid biosynthesis. Mitochondrial C1 metabolism, including serine hydroxymethyltransferase (SHMT) 2, provides glycine, NAD(P)H, ATP, and C1 units for cytosolic biosynthetic reactions
Aamod S Dekhne et al.
Molecular pharmacology, 97(1), 9-22 (2019-11-11)
Folate-dependent one-carbon (C1) metabolism is compartmentalized in the mitochondria and cytosol and is a source of critical metabolites for proliferating tumors. Mitochondrial C1 metabolism including serine hydroxymethyltransferase 2 (SHMT2) generates glycine for de novo purine nucleotide and glutathione biosynthesis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service