Skip to Content
Merck
All Photos(1)

Key Documents

SML1659

Sigma-Aldrich

NCT-503

≥98% (HPLC)

Synonym(s):

N-(4,6-Dimethylpyridin-2-yl)-4-(4-(trifluoromethyl)benzyl)piperazine-1-carbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23F3N4S
CAS Number:
Molecular Weight:
408.48
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC(NC(N2CCN(CC3=CC=C(C(F)(F)F)C=C3)CC2)=S)=NC(C)=C1

InChI

1S/C20H23F3N4S/c1-14-11-15(2)24-18(12-14)25-19(28)27-9-7-26(8-10-27)13-16-3-5-17(6-4-16)20(21,22)23/h3-6,11-12H,7-10,13H2,1-2H3,(H,24,25,28)

InChI key

PJNSZIQUFLWRLH-UHFFFAOYSA-N

Biochem/physiol Actions

NCT-503 is an inhibitor of phosphoglycerate dehydrogenase (PHGDH), which catalyzes the first, rate-limiting step of glucose-derived serine synthesis. NCT-503 reduced glucose-derived serine production and suppressed the growth of both PHGDH-dependent cancer cells in culture and in xenograft tumors, and caused G1/S cell cycle arrest in MDA-MB-468 cells. NCT-503 reduced the incorporation of one-carbon units from glucose-derived and exogenous serine into nucleotides, which may contribute to its anticancer activity. NCT-503 had an IC50 value of 2.5 μM for PHGDH and was inactive against a panel of other dehydrogenases and minimal cross-reactivity in a panel of 168 G-protein-coupled receptors (GPCRs). NCT-503 was found to be noncompetitive with respect to both 3-PG and NAD+, and exhibit good stability (>98% after 48 hrs in assay buffer) and aqueous solubility.

Recommended products

NCT-503 Inactive Control, a structurally similar analog of NCT-503, is available from Sigma. To learn more about and purchase NCT-503 inactive control, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nirmalya Sen et al.
Molecular carcinogenesis, 57(10), 1342-1357 (2018-06-07)
Ewing sarcoma (EWS) is a soft tissue and bone tumor that occurs primarily in adolescents and young adults. In most cases of EWS, the chimeric transcription factor, EWS-FLI1 is the primary oncogenic driver. The epigenome of EWS cells reflects EWS-FLI1
Samah Elsaadi et al.
Experimental hematology & oncology, 10(1), 3-3 (2021-01-06)
Multiple myeloma (MM) is a hematological malignancy characterized by the clonal expansion of plasma cells in the bone marrow. To date, this disease is still incurable and novel therapeutic approaches are required. Phosphoglycerate dehydrogenase (PHGDH) is the first and rate-limiting
Yingfeng Xia et al.
Cancer research, 79(15), 3837-3850 (2019-05-16)
MYCN amplification drives the development of neuronal cancers in children and adults. Given the challenge in therapeutically targeting MYCN directly, we searched for MYCN-activated metabolic pathways as potential drug targets. Here we report that neuroblastoma cells with MYCN amplification show
Birte Arlt et al.
Journal of enzyme inhibition and medicinal chemistry, 36(1), 1282-1289 (2021-07-02)
The small-molecule inhibitor of phosphoglycerate dehydrogenase, NCT-503, reduces incorporation of glucose-derived carbons into serine in vitro. Here we describe an off-target effect of NCT-503 in neuroblastoma cell lines expressing divergent phosphoglycerate dehydrogenase (PHGDH) levels and single-cell clones with CRISPR-Cas9-directed PHGDH
Shauni Lien Geeraerts et al.
Molecular cancer therapeutics, 20(1), 50-63 (2020-11-19)
Metabolic rewiring is a hallmark of cancer that supports tumor growth, survival, and chemotherapy resistance. Although normal cells often rely on extracellular serine and glycine supply, a significant subset of cancers becomes addicted to intracellular serine/glycine synthesis, offering an attractive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service