Skip to Content
Merck
All Photos(1)

Key Documents

140325

Sigma-Aldrich

8-Hydroxy-5-nitroquinoline

96%

Synonym(s):

5-Nitro-8-quinolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6N2O3
CAS Number:
Molecular Weight:
190.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

powder

mp

181-183 °C (lit.)

solubility

alcohol: very slightly soluble
diethyl ether: very slightly soluble
hydrochloric acid: freely soluble (hot)

functional group

nitro

SMILES string

Oc1ccc([N+]([O-])=O)c2cccnc12

InChI

1S/C9H6N2O3/c12-8-4-3-7(11(13)14)6-2-1-5-10-9(6)8/h1-5,12H

InChI key

RJIWZDNTCBHXAL-UHFFFAOYSA-N

Application

8-Hydroxy-5-nitroquinoline was used in the synthesis of novel CO2-soluble 8-hydroxyquinoline chelating agents.

Biochem/physiol Actions

8-Hydroxy-5-nitroquinoline is an effective anti-microbial and anti-cancer agent. It is an effective drug for the treatment of urinary tract infections due to gram negative bacilli.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gau-Yi Gao et al.
Journal of pharmaceutical sciences, 99(1), 255-261 (2009-06-17)
To investigate the physicochemical characteristics, thermodynamics, possible sublimation process and kinetics of nitroxoline, differential scanning calorimetry (DSC), isothermal thermogravimetry (TG), and Fourier transform infrared (FTIR) microspectroscopy equipped with a micro hot-stage of DSC microscopy assembly (simultaneous DSC-FTIR method) were used.
A V Yatsenko et al.
Acta crystallographica. Section C, Crystal structure communications, 58(Pt 1), o19-o21 (2002-01-10)
The crystal structures of 8-hydroxy-5-nitroquinoline, C9H6N2O3, (I), and 8-hydroxy-5-nitroquinolinium chloride, C9H7N2O3+*Cl-, (II), have been determined from X-ray powder data. In (I), the molecules are linked via moderately strong hydrogen bonds to form dimers. Such a packing motif is likely to
C Pelletier et al.
Antimicrobial agents and chemotherapy, 39(3), 707-713 (1995-03-01)
The antibacterial activity of nitroxoline (NIT), an antibiotic used in the treatment of acute or recurrent urinary tract infections caused by Escherichia coli, is decreased in the presence of Mg2+ and Mn2+ but not Ca2+. In order to elucidate the
Mohamed M Ghoneim et al.
Bioelectrochemistry (Amsterdam, Netherlands), 80(2), 162-168 (2010-09-14)
Nitroxoline has been reduced at the mercury electrode in buffered solutions (pH 2-11) in two irreversible cathodic steps. The first step was attributed to reduction of -NO(2) group to the hydroxylamine stage and the second one to reduction-saturation of the
E Bergogne-Berezin et al.
Pathologie-biologie, 35(5 Pt 2), 873-878 (1987-06-01)
Nitroxoline (8-hydroxy-5-nitroquinoline) has been used since 1962 in the treatment of urinary tract infections especially those due to gram negative bacilli (E. coli). The current renewal of the drug is in relation to the recently shown activity of nitroxoline against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service