Skip to Content
Merck
All Photos(1)

Key Documents

SML2572

Sigma-Aldrich

AZM475271

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27ClN4O3
CAS Number:
Molecular Weight:
442.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC=C(C=C1NC2=NC=NC3=CC(OCC4CCN(CC4)C)=C(C=C32)OC)OC

InChI

1S/C23H27ClN4O3/c1-28-8-6-15(7-9-28)13-31-22-12-19-17(11-21(22)30-3)23(26-14-25-19)27-20-10-16(29-2)4-5-18(20)24/h4-5,10-12,14-15H,6-9,13H2,1-3H3,(H,25,26,27)

InChI key

WPOXAFXHRJYEIC-UHFFFAOYSA-N

Biochem/physiol Actions

AZM475271 (M475271) is an orally active, potent and selective inhibitor against src family kinases src and yes (IC50 = 25 nM & 10 nM, respectively; lck/VEGFR2/EGFR/csk/FGFR1 IC50 = 0.2/0.5/0.6/7.6/∼20 μM) that effectively suppresses src-dependent cellular signaling events (26%/48% inhibition of 50 ng/mL VEGF-induced HUVEC proliferation/migration at 100 nM; 31%/72% inhibition of 10 ng/mL VEGF-induced Flk-1/Src phosphorylation in HUVECs at 300 nM) and exbihits anticancer efficacy both in cultures (70% PC-9/30% A549 proliferation inhibition at 1 μM) and in vivo (78%/100% PC-9 tumor growth suppression in mice via 10/50 mg/kg/day p.o.).
Orally active, potent and selective src and yes inhibitor with anticancer efficacy both in cultures and in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suhrid Banskota et al.
PloS one, 11(1), e0148133-e0148133 (2016-01-30)
5-Hydroxytryptamine (5-HT) induces proliferation of cancer cells and vascular cells. In addition to 5-HT production by several cancer cells including gastrointestinal and breast cancer, a significant level of 5-HT is released from activated platelets in the thrombotic environment of tumors
Yacine Merrouche et al.
Oncotarget, 7(33), 53350-53361 (2016-07-28)
Estrogen receptor-, progesterone receptor- and HER2-negative breast cancers, also known as triple-negative breast cancers (TNBCs), have poor prognoses and are refractory to current therapeutic agents, including epidermal growth factor receptor (EGFR) inhibitors. Resistance to anti-EGFR therapeutic agents is often associated
Rui Zheng et al.
Clinical & experimental metastasis, 22(3), 195-204 (2005-09-15)
Src, a proto-oncogene, has been strongly implicated in the growth, progression and metastasis of a number of human cancers. Its role in lung cancer is, however, still unknown. In the present study, we assessed the expression of Src in three
Nermin Ali et al.
Journal of pharmacological sciences, 98(2), 130-141 (2005-06-07)
Vascular endothelial growth factor (VEGF) was reported to be a potent proangiogenic factor that plays a pivotal role in both physiological and pathological angiogenesis. M475271, 4-quinazolinamine, N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl) methoxy]-(9Cl), is a new anilinoquinazoline derivative that showed selective inhibition of Src kinase
Jaya Gautam et al.
Molecular cancer, 15(1), 75-75 (2016-11-23)
Triple-negative breast cancer (TNBC) has a high risk of relapse and there are few chemotherapy options. Although 5-hydroxytryptamine (5-HT, serotonin) signaling pathways have been suggested as potential targets for anti-cancer drug development, the mechanism responsible for the action of 5-HT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service