Skip to Content
Merck
All Photos(1)

Key Documents

SML1923

Sigma-Aldrich

A-395 hydrochloride

≥98% (HPLC)

Synonym(s):

(3R,4S)-1-(7-Fluoro-2,3-dihydro-1H-inden-1-yl)-4-{4-[4-(methanesulfonyl)piperazin-1-yl]phenyl}-N,N-dimethylpyrrolidin-3-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35FN4O2S · xHCl
CAS Number:
Molecular Weight:
486.65 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=CC2=C1C(N3C[C@H](C(C=C4)=CC=C4N5CCN(S(C)(=O)=O)CC5)[C@@H](N(C)C)C3)CC2

InChI

1S/C26H35FN4O2S/c1-28(2)25-18-30(24-12-9-20-5-4-6-23(27)26(20)24)17-22(25)19-7-10-21(11-8-19)29-13-15-31(16-14-29)34(3,32)33/h4-8,10-11,22,24-25H,9,12-18H2,1-3H3/t22-,24?,25+/m1/s1

InChI key

REVJNSVNICWODC-KIDMSAQOSA-N

Biochem/physiol Actions

A-395 is a potent and selective chemical probe for the polycomb protein EED (embryonic ectoderm development), an essential component of Polycomb repressive complex 2 (PRC2), involved in transcriptional repression through methylation of histone H3K27. A-395 has been found to bind to EED in vitro with a Ki value of 0.4 nM, inhibit the PRC2 complex with an IC50 value 34 nM for methylation of H3K27, and have >100-fold selectivity over other histone methyltransferases and non-epigenetic targets. In RD rhabdoid tumor cell line A-395 inhibited the PRC2 complex with an IC50 value of 90 nM, inhibiting the formation of H3K27me3. For characterization details of A-395, please visit the A-395 probe summary on the Structural Genomics Consortium (SGC) website.

A-395N is the negative control for the active enantiomer, A-395. A-395N is available from Sigma. To learn more about and purchase A-395N, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
Potent and selective chemical probe for EED

Features and Benefits

A-395 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Yang et al.
eLife, 12 (2023-09-12)
Human health is facing a host of new threats linked to unbalanced diets, including high-sugar diet (HSD), which contributes to the development of both metabolic and behavioral disorders. Studies have shown that diet-induced metabolic dysfunctions can be transmitted to multiple
Coral K Wille et al.
Science advances, 9(46), eadf3980-eadf3980 (2023-11-17)
Embryonic stem cells (ESCs) have transcriptionally permissive chromatin enriched for gene activation-associated histone modifications. A striking exception is DOT1L-mediated H3K79 dimethylation (H3K79me2) that is considered a positive regulator of transcription. We find that ESCs are depleted for H3K79me2 at shared
Hywel Dunn-Davies et al.
Molecular therapy. Nucleic acids, 35(2), 102173-102173 (2024-04-15)
Epigenetic processes involving long non-coding RNAs regulate endothelial gene expression. However, the underlying regulatory mechanisms causing endothelial dysfunction remain to be elucidated. Enhancer of zeste homolog 2 (EZH2) is an important rheostat of histone H3K27 trimethylation (H3K27me3) that represses endothelial
Anne-Marie W Turner et al.
ACS infectious diseases, 6(7), 1719-1733 (2020-04-30)
A hallmark of human immunodeficiency type-1 (HIV) infection is the integration of the viral genome into host chromatin, resulting in a latent reservoir that persists despite antiviral therapy or immune response. Thus, key priorities toward eradication of HIV infection are
Alastair Davies et al.
Nature cell biology, 23(9), 1023-1034 (2021-09-08)
Cancers adapt to increasingly potent targeted therapies by reprogramming their phenotype. Here we investigated such a phenomenon in prostate cancer, in which tumours can escape epithelial lineage confinement and transition to a high-plasticity state as an adaptive response to potent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service