Skip to Content
Merck
All Photos(1)

Documents

SML1832

Sigma-Aldrich

DDR1 Inhibitor 7rh

≥98% (HPLC)

Synonym(s):

4-Ethyl-N-(3-((4-methylpiperazin-1-yl)methyl)-5-(trifluoromethyl)-phenyl)-3-(pyrazolo[1,5-a]pyrimidin-6-ylethynyl)benzamide, 7rh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H29F3N6O
CAS Number:
Molecular Weight:
546.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C(F)(F)F)=CC(CN2CCN(C)CC2)=C1)C3=CC=C(CC)C(C#CC4=CN5C(N=C4)=CC=N5)=C3

General description

DDR1 Inhibitor 7rh is a small potent kinase inhibitor of discoidin domain receptor-1 (DDR1). It inhibits the tumorigenicity of nasopharyngeal carcinoma (NPC) cells. DDR1 Inhibitor 7rh prevents the proliferation, invasion and adhesion of cancer cells.

Biochem/physiol Actions

DDR1 Inhibitor 7rh is an orally available, potent, ATP-competitive DDR1-selective inhibitor with in vitro and in vivo anti-cancer efficacy.
Compound 7rh is a potent, high affinity (Kd =0.6 nM), ATP-competitive inhibitor against discoidin domain-containing receptor 1 (DDR1; IC50 = 6.8 nM, [ATP] = 100 nM) with significantly reduced potency toward 455 other kinases, including DDR2, Bcr-abl, and c-Kit (IC50 = 101.4 nM, 355 nM and >10 μM, respectively). Inhibitor 7rh reduces DDR1 expression/phosphorylation and downstream signaling in a dose-dependent manner (0.1-2 μM; NCI-H23 NSCLCs), effectively suppressing human cancer cells proliferation (IC50 from 38 nM/K562 to 2.98 μM/NCI-H460) and colony formation (IC50 = 0.56 μM/NCI-H23). Inhibitor 7rh is orally availabe in rats and mice (T1/2 = 15.53 h; Tmax = 4.25 h; Cmax = 1867.5 μg/L, F = 67.4%; 25 mg/kg; rats) and displays in vivo efficacy against Kras (LSLG12Vgeo) tumor growth in mice (50 mg/kg/day p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingshan Gao et al.
Journal of medicinal chemistry, 56(8), 3281-3295 (2013-03-26)
Discoidin domain receptor 1 (DDR1) is an emerging potential molecular target for new anticancer drug discovery. We have discovered a series of 3-(2-(pyrazolo[1,5-a]pyrimidin-6-yl) ethynyl)benzamides that are selective and orally bioavailable DDR1 inhibitors. The two most promising compounds (7rh and 7rj)
Chiara Ambrogio et al.
Nature medicine, 22(3), 270-277 (2016-02-09)
Patients with advanced Kirsten rat sarcoma viral oncogene homolog (KRAS)-mutant lung adenocarcinoma are currently treated with standard chemotherapy because of a lack of efficacious targeted therapies. We reasoned that the identification of mediators of Kras signaling in early mouse lung
Antitumor activity of 7RH, a discoidin domain receptor 1 inhibitor, alone or in combination with dasatinib exhibits antitumor effects in nasopharyngeal carcinoma cells
Lu QP, et al.
Oncology Letters, 12(5), 3598-3608 (2016)
Protein Analysis and Purification: Benchtop Techniques, 3598-3608 (2018)
Hoon Hur et al.
BMC cancer, 17(1), 87-87 (2017-02-02)
Discoidin domain receptor 1 (DDR1), a receptor tyrosine kinase that utilizes collagen as a ligand, is a key molecule in the progression of solid tumors as it regulates the interaction of cancer cells with the tumor stroma. However, the clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service