Skip to Content
Merck
All Photos(1)

Documents

CDS022173

Sigma-Aldrich

Imatinib

Synonym(s):

4-(4-Methylpiperazin-1-ylmethyl)-N-[4-methyl-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]phenyl]benzamide, 4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, CGP 57148, Genfatinib, N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31N7O
CAS Number:
Molecular Weight:
493.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

O=C(C(C=C1)=CC=C1CN2CCN(C)CC2)NC3=CC(NC4=NC(C5=CN=CC=C5)=CC=N4)=C(C)C=C3

InChI

1S/C29H31N7O/c1-21-5-10-25(18-27(21)34-29-31-13-11-26(33-29)24-4-3-12-30-19-24)32-28(37)23-8-6-22(7-9-23)20-36-16-14-35(2)15-17-36/h3-13,18-19H,14-17,20H2,1-2H3,(H,32,37)(H,31,33,34)

InChI key

KTUFNOKKBVMGRW-UHFFFAOYSA-N

Gene Information

General description

Imatinib is a benzamide derivative used as a is specific tyrosine kinase receptor inhibitor. It is designed to inhibit the breakpoint cluster region (BCR)-Abelson (ABL) fusion protein, which is caused by the Philadelphia chromosome abnormalities. Imatinib is also used as a potent apoptosis inducer and an antineoplastic agent.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent L Reber et al.
PloS one, 12(10), e0185704-e0185704 (2017-10-06)
Gouty arthritis is caused by the deposition of monosodium urate (MSU) crystals in joints. Despite many treatment options for gout, there is a substantial need for alternative treatments for patients unresponsive to current therapies. Tyrosine kinase inhibitors have demonstrated therapeutic
Xiaohui Si et al.
Oncotarget, 7(47), 78095-78109 (2016-11-02)
Leukemia stem cells (LSCs) can resist available treatments that results in disease progression and/or relapse. To dissect the microRNA (miRNA) expression signature of relapse in acute myeloid leukemia (AML), miRNA array analysis was performed using enriched LSCs from paired bone
Tamalika Paul et al.
Anti-cancer drugs, 30(2), 167-178 (2018-11-13)
The tumor necrosis factor-related apoptosis-inducing ligand (TRAIL), a member of cytokine superfamily, induces apoptosis in a number of tumor cells through the activation of extrinsic apoptotic pathway but shows little or no cytotoxicity toward normal cells. However some tumor cells
Bożena Karolewicz et al.
Pharmaceutics, 11(7) (2019-07-04)
The proposed study examined the characterization and stability of solid-state amorphous imatinib mesylate (IM) after 15 months under controlled relative humidity (60 ± 5%) and temperature (25 ± 2 °C) conditions. After 2 weeks, and 1, 3, 6, and 15
Doing evidence-based medicine? How NHS managers ration high-cost drugs.
David Hughes et al.
Social science & medicine (1982), 235, 112304-112304 (2019-07-16)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service