Skip to Content
Merck
All Photos(1)

Key Documents

SML0632

Sigma-Aldrich

Fialuridine

≥98% (HPLC), powder, nucleoside analog

Synonym(s):

1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-iodo-2,4(1H,3H)-Pyrimidinedione, FIAU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10FIN2O5
CAS Number:
Molecular Weight:
372.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Fialuridine, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

F[C@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C(NC(C(I)=C2)=O)=O

InChI

1S/C9H10FIN2O5/c10-5-6(15)4(2-14)18-8(5)13-1-3(11)7(16)12-9(13)17/h1,4-6,8,14-15H,2H2,(H,12,16,17)/t4-,5+,6-,8-/m1/s1

InChI key

IPVFGAYTKQKGBM-BYPJNBLXSA-N

General description

Fialuridine (1-(2-deoxy-2-fluoro-β-d-arabinofuranosyl)-5-iodouracil, or FIAU) is an antiviral agent. It is a thymidine-based nucleoside analogue.

Application

Fialuridine has been used in the selection of clones.

Biochem/physiol Actions

Fialuridine (1-(2-deoxy-2-fluoro-β-d-arabinofuranosyl)-5-iodouracil, or FIAU) and its metabolites blocks DNA polymerase at sites of multiple adjacent analog incorporation, reduces the presence of mtDNA (mitochondrial DNA) and results in mitochondrial structural defects in cultured hepatoblasts. It is considered as an efficient drug against hepatitis B virus (HBV) infection.
Fialuridine is a nucleoside analog antiviral agent.
Fialuridine is a nucleoside analog antiviral agent. The compound displays significant mitochondrial toxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

P Honkoop et al.
Drug safety, 17(1), 1-7 (1997-07-01)
Fialuridine is an antiviral agent with potent activity against hepatitis B virus replication in vitro and in vivo. In a phase II study, 7 of 15 patients experienced severe toxicity due to the drug after 9 to 13 weeks of
Kurt M Lin et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 10(5), 253-263 (2008-06-19)
Human ZR75-1 cells were among the first few characterized estrogen-dependent mammary gland carcinoma cell lines and had been utilized in various studies for the pro- or antitumor effect of xenoestrogens and antiestrogens. The objective of this study was to establish
Hepatic failure and lactic acidosis due to fialuridine (FIAU), an investigational nucleoside analogue for chronic hepatitis B.
McKenzie R, et al.
The New England Journal of Medicine, 333(17), 1099-1105 (1995)
Fialuridine accumulates in DNA of dogs, monkeys, and rats following long-term oral administration.
Richardson FC, et al.
Proceedings of the National Academy of Sciences of the USA, 91(25), 12003-12007 (1994)
Fialuridine and its metabolites inhibit DNA polymerase gamma at sites of multiple adjacent analog incorporation, decrease mtDNA abundance, and cause mitochondrial structural defects in cultured hepatoblasts.
Lewis W, et al.
Proceedings of the National Academy of Sciences of the USA, 93(8), 3592-3597 (1996)

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service