Skip to Content
Merck
All Photos(1)

Key Documents

SML0468

Sigma-Aldrich

Dorzolamide

≥98% (HPLC)

Synonym(s):

(4S,6S)-4H-Thieno[2,3-b]thiopyran-2-sulfonamide, 4-(ethylamino)-5,6-dihydro-6-methyl-, 7,7-dioxide, 4S,6S-Dorzolamide, L-671,152, MK-507

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O4S3
CAS Number:
Molecular Weight:
324.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -25 to -35°, c = 1 in methanol

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution, warmed)

storage temp.

room temp

SMILES string

CCN[C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(N)(=O)=O

InChI

1S/C10H16N2O4S3/c1-3-12-8-4-6(2)18(13,14)10-7(8)5-9(17-10)19(11,15)16/h5-6,8,12H,3-4H2,1-2H3,(H2,11,15,16)/t6-,8-/m0/s1

InChI key

IAVUPMFITXYVAF-XPUUQOCRSA-N

Gene Information

human ... CA2(760)

Biochem/physiol Actions

Dorzolamide is a potent carbonic anhydrase II inhibitor with an IC50 value of 0.16 nM on human erythrocyte carbonic anhydrase II in vitro. It has been found to lower increased intraocular pressure in open-angle glaucoma and ocular hypertension.
Dorzolamide is a potent carbonic anhydrase II inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ichiro Kawaguchi et al.
PloS one, 12(10), e0186714-e0186714 (2017-10-20)
This study aimed to determine if rebamipide eyedrops can improve ocular surface damage caused by the use of glaucoma eyedrops. Female Kbl:Dutch rabbits were used to evaluate glaucoma eyedrop-induced ocular surface damage; one eye of each rabbit was untreated and
Luciano Quaranta et al.
Journal of glaucoma, 26(11), 987-994 (2017-09-29)
To investigate the efficacy and safety of a treatment strategy with latanoprost and dorzolamide in primary pediatric glaucoma patients partially responsive to surgery. Children with primary pediatric glaucoma having postsurgical untreated intraocular pressure (IOP) between 22 and 26 mm Hg were eligible.
Riku P J Arvola et al.
Cornea, 35(4), 451-455 (2016-02-05)
Treating corneal neurotrophic ulcers is challenging. Topical application of matrix regeneration therapy (RGTA), which is a dextran derivative polymer and heparan sulfate analog, is a promising regenerative therapy and an alternative or additional therapeutic regimen when corneal healing is compromised.
Yukari Yagi-Yaguchi et al.
Diagnostics (Basel, Switzerland), 10(1) (2020-01-08)
Anti-glaucoma eye drop treatment often induces dry eyes and can lead to poor medication adherence. This study aimed to investigate the effects of 3% diquafosol sodium eye drops on tear function and the ocular surface epithelium in Sod1-/- mice after
Frank Bochmann et al.
Journal of glaucoma, 26(11), 1051-1055 (2017-10-28)
A subluxation or dislocation of the intraocular lens (IOL) is a well-known complication after cataract surgery in patients with pseudoexfoliation syndrome (PEX). Capsular tension rings (CTR) are frequently used to prevent postoperative complications caused by zonular weakness. Here we present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service