Skip to Content
Merck
All Photos(1)

Key Documents

SMB01014

Sigma-Aldrich

Clemastanin B

≥90% (LC/MS-ELSD)

Synonym(s):

LAriciresinol 4,4′-bis-O-|A-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H44O16
CAS Number:
Molecular Weight:
684.68
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

684.68

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]([C@H]([C@H](C1)Cc4cc(c(cc4)O[C@@H]5O[C@@H]([C@H]([C@@H]([C@H]5O)O)O)CO)OC)CO)c2cc(c(cc2)O[C@@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)OC

InChI

1S/C32H44O16/c1-42-20-8-14(3-5-18(20)45-31-28(40)26(38)24(36)22(11-34)47-31)7-16-13-44-30(17(16)10-33)15-4-6-19(21(9-15)43-2)46-32-29(41)27(39)25(37)23(12-35)48-32/h3-6,8-9,16-17,22-41H,7,10-13H2,1-2H3/t16-,17-,22+,23+,24+,25+,26-,27-,28+,29+,30+,31+,32+/

InChI key

PBLWZMSRSJTRHJ-NCIRKIHRSA-N

General description

Clemastanin B, a furanoid lignan, is commonly produced by Clematis stans, Radix Isatidis, Plocama calabrica, Sedum sarmentosum, and Brassica rapa plants. Existing research indicates that this secondary metabolite, derived from plant source, exhibits various biological properties, including antiviral, antiinflammatory, and antioxidant activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Clemastanin B has antioxidant and anti-inflammatory activities, it exerts its anti-influenza activity by inhibiting the virus multiplication, prophylaxsis and blocking the virus attachment.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Xiao et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 36(3), 369-376 (2016-07-30)
To study the antiviral activities of clemastanin B (CB), epigoitrin, phenylpropanoids portion (PEP) and the mixture of phenylpropanoids, alkaloids and organic acid fractions (PEP+ALK+OA) from Banlangen (Radix Isatidis). The experiment consisted of four parts: therapeutic action, prophylaxsis action, inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service