Skip to Content
Merck
All Photos(1)

Key Documents

40674

Sigma-Aldrich

Pinoresinol

≥95.0% (HPLC)

Synonym(s):

(+)-Pinoresinol, 4,4′-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol), 4,4′-[(1S,3aR,4S,6aR)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2-methoxyphenol)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95.0% (HPLC)

form

powder or crystals

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

COc1cc(ccc1O)[C@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4ccc(O)c(OC)c4

InChI

1S/C20H22O6/c1-23-17-7-11(3-5-15(17)21)19-13-9-26-20(14(13)10-25-19)12-4-6-16(22)18(8-12)24-2/h3-8,13-14,19-22H,9-10H2,1-2H3/t13-,14-,19+,20+/m0/s1

InChI key

HGXBRUKMWQGOIE-AFHBHXEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pinoresinol is a secondary metabolite, a lignan found in wide varieties of plants. Structurally, it is one of the simple lignans, with a dimer of coniferyl alcohol, which forms the bicyclic ring core.

Application

Pinoresinol has been used:
  • as a reference standard for qualitative and quantitative analysis of lignans from Triticale (X Triticosecale Wittmack) grains using ultra-performance liquid chromatography (UPLC) with photodiode and mass TQD detectors
  • as an enterolignan precursor to study its estrogenic activity on the proliferation of human breast cancer MCF-7 cells
  • as a reference standard for lignan analysis of Sesamum indicum L. seeds

Biochem/physiol Actions

Pinoresinol is present in a number of plants including medical plants like Sambucus williamsii, Eucommia ulmoides (duzhong), Styrax sp, Forsythia suspensa, and in extra virgin olive oil. The genus Sambucus is widely distributed in Europe, Asia, and North Africa, and has been used in traditional medicine as an analgesic, antivirus, antiinflammatory, homoeostatic, and diuretic drugs which act on bruises, fractures, and edema. Pinoresinol displays potent antifungal properties by causing damage to the fungal plasma membrane. It exerts anti-oxidative and anti-inflammatory activities by inhibition of TNF- α production (presumably through inhibition of NF- κB and AP-1).
exerts anti-oxidative and anti-inflammatory activities by inhibition of TNF-α production

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kishan Chandra et al.
Biotechnology reports (Amsterdam, Netherlands), 22, e00336-e00336 (2019-04-25)
Members of Cytochromes P450 super family of enzymes catalyse important biochemical reactions in plants. Some of these reactions are so important that they contribute to enormous chemical diversity seen in plants. Many unique secondary metabolites formed by mediation of these
Frank C Schroeder et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(42), 15497-15501 (2006-10-13)
Pinoresinol, a lignan of wide distribution in plants, is found to occur as a minor component in the defensive secretion produced by glandular hairs of caterpillars of the cabbage butterfly, Pieris rapae. The compound or a derivative is appropriated by
Lignans in triticale grain and triticale products
Makowska A, et al.,
Journal of Cereal Science, 93, 102939-102939 (2020)
Yun Zhu et al.
PloS one, 12(2), e0171390-e0171390 (2017-02-06)
Mammalian lignans or enterolignans are metabolites of plant lignans, an important category of phytochemicals. Although they are known to be associated with estrogenic activity, cell signaling pathways leading to specific cell functions, and especially the differences among lignans, have not
Hyo Won Jung et al.
Neuroscience letters, 480(3), 215-220 (2010-07-06)
The activation of microglia plays an important role in a variety of brain disorders by the excessive production of inflammatory mediators such as nitric oxide (NO), prostaglandin E(2) (PGE(2)) and proinflammatory cytokines. We investigated here whether pinoresinol isolated from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service