Skip to Content
Merck
All Photos(1)

Key Documents

SMB00532

Sigma-Aldrich

Amorfrutin B

≥85% (NMR)

Synonym(s):

3-(3,7-dimethyloctyl)-2-hydroxy-4-methoxy-6-(2-phenylethyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32O4
CAS Number:
Molecular Weight:
408.53
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥85% (NMR)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(C)=CCC/C(C)=C/CC1=C(OC)C=C(CCC2=CC=CC=C2)C(C(O)=O)=C1O

InChI

1S/C26H32O4/c1-18(2)9-8-10-19(3)13-16-22-23(30-4)17-21(24(25(22)27)26(28)29)15-14-20-11-6-5-7-12-20/h5-7,9,11-13,17,27H,8,10,14-16H2,1-4H3,(H,28,29)/b19-13+

InChI key

DKNIJLWIVUCTHW-CPNJWEJPSA-N

General description

Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chu Chen et al.
Pharmaceutical biology, 54(3), 488-493 (2015-05-26)
Initially isolated from fruits of Amorpha fruticosa L. (Fabaceae), amorfrutins are promising antidiabetic natural products as selective peroxisome proliferator-activated receptor γ-agonists. The objective of this study is to develop a sensitive and convenient HPLC method to analysis amorfrutins in plant
Chu Chen et al.
Journal of separation science, 38(16), 2924-2929 (2015-06-05)
Prenylated phenolics such as amorfrutins are recently identified potent anti-inflammatory and antidiabetic natural products. In this work, high-speed counter-current chromatography was investigated for the isolation and purification of prenylated phenolics from the fruits of Amorpha fruticosa by using a two-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service