Skip to Content
Merck
All Photos(1)

Key Documents

SBR00002

Sigma-Aldrich

Arzanol

from Helichrysum italicum, ≥98%

Synonym(s):

3-[[3-Acetyl-2,4,6-trihydroxy-5-(3-methyl-2-buten-1-yl)phenyl]methyl]-6-ethyl-4-hydroxy-5-methyl-2H-pyran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26O7
CAS Number:
Molecular Weight:
402.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

biological source

Helichrysum italicum

Quality Level

Assay

≥98%

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=C(CC2=C(O)C(C)=C(CC)OC2=O)C(O)=C(C(C)=O)C(O)=C1CC=C(C)C

InChI

1S/C22H26O7/c1-6-16-11(4)18(24)15(22(28)29-16)9-14-19(25)13(8-7-10(2)3)20(26)17(12(5)23)21(14)27/h7,24-27H,6,8-9H2,1-5H3

InChI key

ZOIAPLVBZQQHCG-UHFFFAOYSA-N

General description

Arzanol is a pyrone–phloroglucinol etherodimer. It is a polyphenol compound extracted from the plant Helichrysum italicum which grows in the Mediterranean area.

Application

Arzanol has been used to test its effects as an autophagy modulator on HeLa cells and bladder cancer cells

Biochem/physiol Actions

Arzanol is a polyphenol compound extracted from the plant Helichrysum italicum which grows in the Mediterranean area. Arzanol is known for its anti-inflammatory and antimicrobial activities. The anti-inflammatory activity was verified in an in-vivo rat model and was found to suppress the inflammatory response of the carrageenan-induced pleurisy. Moreover it was found to have an anti-HIV-1 activity. Arzanol inhibits COX-2-derived prostaglandin synthase-1 (mPGES-1) Inhibitor in-vitro, in human stimulated monocytes.
Arzanol inhibits the activation of inflammatory transcription factor NFκB, HIV replication in T cells, releases of IL-1β, IL-6, IL-8, and TNF-α,and biosynthesis of PGE2 by potentially inhibiting the mPGES-1 enzyme.
Arzanol is known for its anti-inflammatory and antimicrobial activities. The anti-inflammatory activity was verified in an in-vivo rat model and was found to suppress the inflammatory response of the carrageenan-induced pleurisy. Moreover it was found to have an anti-HIV-1 activity. Arzanol inhibits COX-2-derived prostaglandin synthase-1 (mPGES-1) Inhibitor in-vitro, in human stimulated monocytes.
Arzanol inhibits the activation of inflammatory transcription factor nuclear factor-KB (NF-KB), human immunodeficiency virus (HIV) replication in T cells, releases of interleukin (IL)-1β, IL-6, IL-8, and tumor necrosis factor (TNF)-α and biosynthesis of prostaglandin E2 (PGE2) by potentially inhibiting the mPGES-1 enzyme. It also possesses antioxidant and cytotoxic activity.

Reconstitution

Soluble in dimethyl sulfoxide (DMSO) at 1 mg/mL; poorly soluble in water

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Deitersen et al.
Cell death & disease, 12(6), 560-560 (2021-06-02)
Autophagy is an intracellular recycling pathway with implications for intracellular homeostasis and cell survival. Its pharmacological modulation can aid chemotherapy by sensitizing cancer cells toward approved drugs and overcoming chemoresistance. Recent translational data on autophagy modulators show promising results in
Evaluation of the antioxidant and cytotoxic activity of arzanol, a prenylated a-pyrone?phloroglucinol etherodimer from Helichrysum italicum subsp. microphyllum.
Rosa A, et al.
Chemico-Biological Interactions, 165(2), 117-126 (2007)
Antonella Rosa et al.
Chemistry and physics of lipids, 164(1), 24-32 (2010-10-12)
This study examines the protective effect of arzanol, a pyrone-phloroglucinol etherodimer from Helichrysum italicum subsp. microphyllum, against the oxidative modification of lipid components induced by Cu(2+) ions in human low density lipoprotein (LDL) and by tert-butyl hydroperoxide (TBH) in cell
Arzanol, a prenylated heterodimeric phloroglucinyl pyrone, inhibits eicosanoid biosynthesis and exhibits anti-inflammatory efficacy in vivo.
Bauer J, et al.
Biochemical Pharmacology, 81(2), 259-268 (2011)
Jana Deitersen et al.
Cell death & disease, 12(6), 560-560 (2021-06-02)
Autophagy is an intracellular recycling pathway with implications for intracellular homeostasis and cell survival. Its pharmacological modulation can aid chemotherapy by sensitizing cancer cells toward approved drugs and overcoming chemoresistance. Recent translational data on autophagy modulators show promising results in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service